missing translation for 'onlineSavingsMsg'
Learn More

2-Butyne-1,4-diol, 99%

CAS: 110-65-6 | C4H6O2 | 86.09 g/mol

Supplier:  Thermo Scientific Chemicals 108080010

 View more versions of this product

Catalog No. AC108080010


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Butyne-1, 4-diol
98.5
54.0°C to 58.0°C
1.2000g/mL
152°C
98.5% min. (GC)
C4H6O2
HOCH2C≡CCH2OH
1 kg
1.2
Solubility in water: 2960g/L (20°C). Other solubilities: soluble in aqueous acids,soluble in alcohol and acetone,insoluble in ether and benzene,soluble in polar solvents
C(C#CCO)O
86.09
CHEBI:16413
99%
110-65-6
100.0
Yellow
238.0°C
Authentic
Plastic bottle
1.4804
MFCD00002915
01, I, 261
2-butyne-1,4-diol, butynediol, 1,4-dihydroxy-2-butyne, 1,4-butynediol, bis hydroxymethyl acetylene, 2-butynediol, 2-butin-1,4-diol, agrisynth b3d, unii-axh202fpqm, 1,4-butynediol van
DLDJFQGPPSQZKI-UHFFFAOYSA-N
but-2-yne-1,4-diol
8066
86.09
Crystalline Platelets or Flakes

Chemical Identifiers

110-65-6
86.09
DLDJFQGPPSQZKI-UHFFFAOYSA-N
8066
but-2-yne-1,4-diol
C4H6O2
MFCD00002915
2-butyne-1,4-diol, butynediol, 1,4-dihydroxy-2-butyne, 1,4-butynediol, bis hydroxymethyl acetylene, 2-butynediol, 2-butin-1,4-diol, agrisynth b3d, unii-axh202fpqm, 1,4-butynediol van
CHEBI:16413
C(C#CCO)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.
Toxic if inhaled.
Causes severe skin burns and eye damage.
May cause damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Toxic in contact w

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 203-788-6

Recommended Storage : Product darkens on storage

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only