Learn More
(2-Carboxyethyl)triphenylphosphonium chloride, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2560922
Description
2-Carboxyethyl)-triphenylphosphonium chloride is a phosphonium ylide reagent for Wittig olefinations with introduction of a carboxylic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Carboxyethyl)-triphenylphosphonium chloride is a phosphonium ylide reagent for Wittig olefinations with introduction of a carboxylic acid.
Solubility
Soluble in dimethyl sulfoxide and methanol.
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
Specifications
(2-Carboxyethyl)triphenylphosphonium chloride | |
197°C to 201°C | |
MFCD00185663 | |
Hygroscopic | |
Soluble in dimethyl sulfoxide and methanol. | |
[Cl-].OC(=O)CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
370.81 | |
370.81 |
36626-29-6 | |
C21H20ClO2P | |
100 g | |
2-carboxyethyl triphenylphosphonium chloride, 2-carboxyethyl tripheylphosphonium chloride, 2-carboxyethyl-triphenylphosphonium chloride, phosphonium, 2-carboxyethyl triphenyl-, chloride, 2-carboxyethyl triphenylphosphanium chloride, acmc-1agne, 2-carboxyethyl triphenyl phosphonium chloride, beta-carboxyethyltriphenylphosphonium chloride, 2-carboxyethyl triphenyl-phosphonium chloride | |
GALLWJZTZYJVSL-UHFFFAOYSA-N | |
2-carboxyethyl(triphenyl)phosphanium;chloride | |
10270534 | |
98% |
Chemical Identifiers
36626-29-6 | |
370.81 | |
GALLWJZTZYJVSL-UHFFFAOYSA-N | |
10270534 |
C21H20ClO2P | |
MFCD00185663 | |
2-carboxyethyl triphenylphosphonium chloride, 2-carboxyethyl tripheylphosphonium chloride, 2-carboxyethyl-triphenylphosphonium chloride, phosphonium, 2-carboxyethyl triphenyl-, chloride, 2-carboxyethyl triphenylphosphanium chloride, acmc-1agne, 2-carboxyethyl triphenyl phosphonium chloride, beta-carboxyethyltriphenylphosphonium chloride, 2-carboxyethyl triphenyl-phosphonium chloride | |
[Cl-].OC(=O)CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only