missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-1,3-thiazole-5-carboxylic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 101012-12-8
Molecular Formula C4H2ClNO2S
Molecular Weight (g/mol) 163.575
InChI Key HNJOKQPEJIWTRF-UHFFFAOYSA-N
Synonym 2-chlorothiazole-5-carboxylic acid, 2-chloro-5-thiazolecarboxylic acid, 5-thiazolecarboxylicacid, 2-chloro, 5-thiazolecarboxylic acid, 2-chloro, acmc-1brcy, 2-chlorothiazol-5-carboxylic acid, 2-chloro-5-carboxy-1,3-thiazole, 5-thiazolecarboxylicacid,2-chloro, 2-chloro-thiazole-5-carboxylic acid
PubChem CID 1481389
IUPAC Name 2-chloro-1,3-thiazole-5-carboxylic acid
SMILES C1=C(SC(=N1)Cl)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC62901DA
SDS View Documents
Thermo Scientific Chemicals
CC62901DA
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC62901DE
SDS View Documents
Thermo Scientific Chemicals
CC62901DE
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

101012-12-8
163.575
2-chlorothiazole-5-carboxylic acid, 2-chloro-5-thiazolecarboxylic acid, 5-thiazolecarboxylicacid, 2-chloro, 5-thiazolecarboxylic acid, 2-chloro, acmc-1brcy, 2-chlorothiazol-5-carboxylic acid, 2-chloro-5-carboxy-1,3-thiazole, 5-thiazolecarboxylicacid,2-chloro, 2-chloro-thiazole-5-carboxylic acid
2-chloro-1,3-thiazole-5-carboxylic acid
C4H2ClNO2S
HNJOKQPEJIWTRF-UHFFFAOYSA-N
1481389
C1=C(SC(=N1)Cl)C(=O)O

Specifications

101012-12-8
C4H2ClNO2S
2-chlorothiazole-5-carboxylic acid, 2-chloro-5-thiazolecarboxylic acid, 5-thiazolecarboxylicacid, 2-chloro, 5-thiazolecarboxylic acid, 2-chloro, acmc-1brcy, 2-chlorothiazol-5-carboxylic acid, 2-chloro-5-carboxy-1,3-thiazole, 5-thiazolecarboxylicacid,2-chloro, 2-chloro-thiazole-5-carboxylic acid
C1=C(SC(=N1)Cl)C(=O)O
163.575
163.58
2-Chloro-1, 3-thiazole-5-carboxylic acid
97%
1g
HNJOKQPEJIWTRF-UHFFFAOYSA-N
2-chloro-1,3-thiazole-5-carboxylic acid
1481389
97%
Videos
Safety and Handling

Safety and Handling

Warning

SDS
missing translation for 'documents'

missing translation for 'documents'