missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-1-fluoro-4-iodobenzene, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1180814

 View more versions of this product

Catalog No. AAA1180814


Explore available promotions

Description

Description

It employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It employed as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Light Sensitive, store in dark. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Specifications

2-Chloro-1-fluoro-4-iodobenzene
2.02
>110°C (230°F)
1.605
25 g
Slightly soluble in water.
C1=CC(=C(C=C1I)Cl)F
256.443
256.45
156150-67-3
94°C to 95°C (15 mmHg)
C6H3ClFI
MFCD00042210
3-chloro-4-fluoroiodobenzene, 1-chloro-2-fluoro-5-iodobenzene, 2-chloro4-iodofluorobenzene, 3-chloro-4-fluoro-1-iodobenzene, 1-fluoro-2-chloro-4-iodobenzene, benzene, 2-chloro-1-fluoro-4-iodo, benzene,2-chloro-1-fluoro-4-iodo, 2-chloro-1-fluoro-4-iodo-benzene, pubchem3433, acmc-20an6s
OMASDGWBVAVFQZ-UHFFFAOYSA-N
2-chloro-1-fluoro-4-iodobenzene
2724605
98%

Chemical Identifiers

156150-67-3
256.443
OMASDGWBVAVFQZ-UHFFFAOYSA-N
2724605
C1=CC(=C(C=C1I)Cl)F
C6H3ClFI
MFCD00042210
3-chloro-4-fluoroiodobenzene, 1-chloro-2-fluoro-5-iodobenzene, 2-chloro4-iodofluorobenzene, 3-chloro-4-fluoro-1-iodobenzene, 1-fluoro-2-chloro-4-iodobenzene, benzene, 2-chloro-1-fluoro-4-iodo, benzene,2-chloro-1-fluoro-4-iodo, 2-chloro-1-fluoro-4-iodo-benzene, pubchem3433, acmc-20an6s
2-chloro-1-fluoro-4-iodobenzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only