Learn More
2-Chloro-2'-arabino-fluoro-2'-deoxyadenosine, 99+%, Thermo Scientific Chemicals
$319.95 - $319.95
Chemical Identifiers
CAS | 123318-82-1 |
---|---|
Molecular Formula | C10H12ClN5O3 |
Molecular Weight (g/mol) | 285.69 |
MDL Number | MFCD00871077 |
InChI Key | ADXUXRNLBYKGOA-GPGUAWMRNA-N |
Synonym | clofarabine, clolar, evoltra, clofarex, cafda, clofarabina, clofarabinum, clofarabine usan, cl-f-ara-a, c1-f-ara-a |
PubChem CID | 119182 |
ChEBI | CHEBI:681569 |
IUPAC Name | (2R,3R,4S,5R)-5-(6-amino-2-chloropurin-9-yl)-4-fluoro-2-(hydroxymethyl)oxolan-3-ol |
SMILES | NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3Cl)C2=NC=N1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ60927MD
|
Thermo Scientific Chemicals
J60927MD |
250 mg |
Each for $319.95
|
|
|||||
Description
A purine nucleoside anti-metabolite. Toxic to nondividing lymphocytes and monocytes
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- A purine nucleoside anti-metabolite
- Toxic to nondividing lymphocytes and monocytes
Chemical Identifiers
123318-82-1 | |
285.69 | |
ADXUXRNLBYKGOA-GPGUAWMRNA-N | |
119182 | |
(2R,3R,4S,5R)-5-(6-amino-2-chloropurin-9-yl)-4-fluoro-2-(hydroxymethyl)oxolan-3-ol |
C10H12ClN5O3 | |
MFCD00871077 | |
clofarabine, clolar, evoltra, clofarex, cafda, clofarabina, clofarabinum, clofarabine usan, cl-f-ara-a, c1-f-ara-a | |
CHEBI:681569 | |
NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3Cl)C2=NC=N1 |
Specifications
123318-82-1 | |
C10H12ClN5O3 | |
250 mg | |
14,2372 | |
ADXUXRNLBYKGOA-GPGUAWMRNA-N | |
(2R,3R,4S,5R)-5-(6-amino-2-chloropurin-9-yl)-4-fluoro-2-(hydroxymethyl)oxolan-3-ol | |
119182 | |
303.68 | |
Powder |
White | |
MFCD00871077 | |
UN2811 | |
clofarabine, clolar, evoltra, clofarex, cafda, clofarabina, clofarabinum, clofarabine usan, cl-f-ara-a, c1-f-ara-a | |
NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3Cl)C2=NC=N1 | |
285.69 | |
CHEBI:681569 | |
≥99% | |
2-Chloro-2'-arabino-fluoro-2'-deoxyadenosine |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P310-P330-P501c
H301
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECSNumber : UD7473000
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only