missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-2-methylbutane, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1170314

 View more versions of this product

Catalog No. AAA1170314


Explore available promotions

Description

Description

2-Chloro-2-methylbutane is used as a solvent for the synthesis of other compounds and pentane. It is used in organic synthesis, Catalytic agent, Petrochemical additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-2-methylbutane is used as a solvent for the synthesis of other compounds and pentane. It is used in organic synthesis, Catalytic agent, Petrochemical additive.

Solubility
Soluble in ethanol, ethyl ether. Insoluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents. Avoid extremes of temperature and direct sunlight.
Specifications

Specifications

2-Chloro-2-methylbutane
-73°C
85°C to 86°C
C5H11Cl
MFCD00000818
UN1107
tert-amyl chloride, butane, 2-chloro-2-methyl, 2-methyl-2-chlorobutane, tert-pentyl chloride, 1,1-dimethylpropyl chloride, tertiary pentyl chloride, t-pentyl chloride, acmc-209mdc, 2-chloro-2-methyl butane, 2-chloro-2-methyl-butane
CRNIHJHMEQZAAS-UHFFFAOYSA-N
2-chloro-2-methylbutane
61143
95%
594-36-5
0.866
−10°C (14°F)
1.405
25 g
1696930
Soluble in ethanol,ethyl ether. Insoluble in water.
CCC(C)(C)Cl
106.593
106.6

Chemical Identifiers

594-36-5
106.593
CRNIHJHMEQZAAS-UHFFFAOYSA-N
61143
CCC(C)(C)Cl
C5H11Cl
MFCD00000818
tert-amyl chloride, butane, 2-chloro-2-methyl, 2-methyl-2-chlorobutane, tert-pentyl chloride, 1,1-dimethylpropyl chloride, tertiary pentyl chloride, t-pentyl chloride, acmc-209mdc, 2-chloro-2-methyl butane, 2-chloro-2-methyl-butane
2-chloro-2-methylbutane
Safety and Handling

Safety and Handling

GHS H Statement
H225
Highly flammable liquid and vapor.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H225

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMYL CHLORIDES

EINECSNumber : 209-836-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only