missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-3-methoxybenzaldehyde 98.0+%, TCI America™
$145.40 - $417.56
Chemical Identifiers
| CAS | 54881-49-1 |
|---|---|
| Molecular Formula | C8H7ClO2 |
| Molecular Weight (g/mol) | 170.592 |
| MDL Number | MFCD07369787 |
| InChI Key | XKLMLKSXPIITAL-UHFFFAOYSA-N |
| PubChem CID | 12575695 |
| IUPAC Name | 2-chloro-3-methoxybenzaldehyde |
| SMILES | COC1=CC=CC(=C1Cl)C=O |
Chemical Identifiers
| 54881-49-1 | |
| 170.592 | |
| XKLMLKSXPIITAL-UHFFFAOYSA-N | |
| 2-chloro-3-methoxybenzaldehyde |
| C8H7ClO2 | |
| MFCD07369787 | |
| 12575695 | |
| COC1=CC=CC(=C1Cl)C=O |
Specifications
| 54881-49-1 | |
| C8H7ClO2 | |
| 5 g | |
| XKLMLKSXPIITAL-UHFFFAOYSA-N | |
| 2-chloro-3-methoxybenzaldehyde | |
| 12575695 | |
| ≥98.0% (GC) | |
| 2-Chloro-3-methoxybenzaldehyde |
| 53°C | |
| MFCD07369787 | |
| 3077 | |
| COC1=CC=CC(=C1Cl)C=O | |
| 170.592 | |
| 170.59 | |
| Crystalline Powder |
Safety and Handling
TSCA : No