missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-4,6-dimethylpyridine, 98+%, Thermo Scientific Chemicals

$195.40 - $882.04

Chemical Identifiers

CAS 30838-93-8
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.60
MDL Number MFCD08277279
InChI Key COVGXNSRDOYAJD-UHFFFAOYSA-N
Synonym 2-chloro-4,6-dimethyl pyridine, 2-chloro-4,6-dimethyl-pyridine, pyridine, 2-chloro-4,6-dimethyl, pubchem3919, acmc-1agqf, ksc495g6l, aebsf hydrochloride; pefabloc sc
PubChem CID 12508378
IUPAC Name 2-chloro-4,6-dimethylpyridine
SMILES CC1=CC(C)=NC(Cl)=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6473306
View Documents
Thermo Scientific Chemicals
H6473306
5 g
Each for $195.40
 
AAH6473314
View Documents
Thermo Scientific Chemicals
H6473314
25 g
Each for $882.04
 
Description

Description

It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

30838-93-8
141.60
COVGXNSRDOYAJD-UHFFFAOYSA-N
12508378
CC1=CC(C)=NC(Cl)=C1
C7H8ClN
MFCD08277279
2-chloro-4,6-dimethyl pyridine, 2-chloro-4,6-dimethyl-pyridine, pyridine, 2-chloro-4,6-dimethyl, pubchem3919, acmc-1agqf, ksc495g6l, aebsf hydrochloride; pefabloc sc
2-chloro-4,6-dimethylpyridine

Specifications

30838-93-8
MFCD08277279
UN2810
2-chloro-4,6-dimethyl pyridine, 2-chloro-4,6-dimethyl-pyridine, pyridine, 2-chloro-4,6-dimethyl, pubchem3919, acmc-1agqf, ksc495g6l, aebsf hydrochloride; pefabloc sc
COVGXNSRDOYAJD-UHFFFAOYSA-N
2-chloro-4,6-dimethylpyridine
12508378
≥98%
C7H8ClN
5 g
108937
Slightly soluble in water.
CC1=CC(C)=NC(Cl)=C1
141.60
141.6
2-Chloro-4,6-dimethylpyridine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only