missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-fluorophenylacetonitrile, 97%

CAS: 75279-56-0 | C8H5ClFN | 169.583 g/mol

$63.03 - $171.40

Chemical Identifiers

CAS 75279-56-0
Molecular Formula C8H5ClFN
Molecular Weight (g/mol) 169.583
MDL Number MFCD00236029
InChI Key GSMCLMKFBYLWRP-UHFFFAOYSA-N
Synonym 2-chloro-4-fluorophenylacetonitrile, 2-chloro-4-fluorobenzyl cyanide, 2-2-chloro-4-fluorophenyl acetonitrile, 2-chloro-4-fluorobenzylcyanide, benzeneacetonitrile, 2-chloro-4-fluoro, 2-chloro-fluorophenylacetonitrile, 2-chloro-4-fluorophenyl acetonitrile, 2-2-chloro-4-fluorophenyl ethanenitrile, pubchem4251
PubChem CID 2725063
IUPAC Name 2-(2-chloro-4-fluorophenyl)acetonitrile
SMILES C1=CC(=C(C=C1F)Cl)CC#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2313206
View Documents
Thermo Scientific Chemicals
B2313206
5 g
Each for $63.03
Only null left
 
AAB2313214
View Documents
Thermo Scientific Chemicals
B2313214
25 g
Each for $171.40
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

75279-56-0
169.583
GSMCLMKFBYLWRP-UHFFFAOYSA-N
2725063
C1=CC(=C(C=C1F)Cl)CC#N
C8H5ClFN
MFCD00236029
2-chloro-4-fluorophenylacetonitrile, 2-chloro-4-fluorobenzyl cyanide, 2-2-chloro-4-fluorophenyl acetonitrile, 2-chloro-4-fluorobenzylcyanide, benzeneacetonitrile, 2-chloro-4-fluoro, 2-chloro-fluorophenylacetonitrile, 2-chloro-4-fluorophenyl acetonitrile, 2-2-chloro-4-fluorophenyl ethanenitrile, pubchem4251
2-(2-chloro-4-fluorophenyl)acetonitrile

Specifications

75279-56-0
C8H5ClFN
5 g
2-chloro-4-fluorophenylacetonitrile, 2-chloro-4-fluorobenzyl cyanide, 2-2-chloro-4-fluorophenyl acetonitrile, 2-chloro-4-fluorobenzylcyanide, benzeneacetonitrile, 2-chloro-4-fluoro, 2-chloro-fluorophenylacetonitrile, 2-chloro-4-fluorophenyl acetonitrile, 2-2-chloro-4-fluorophenyl ethanenitrile, pubchem4251
C1=CC(=C(C=C1F)Cl)CC#N
169.583
169.59
2-Chloro-4-fluorophenylacetonitrile
67°C to 69°C
MFCD00236029
UN3439
GSMCLMKFBYLWRP-UHFFFAOYSA-N
2-(2-chloro-4-fluorophenyl)acetonitrile
2725063
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H311-H331-H315-H319
Fatal if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only