missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-fluorothiophenol, 97%

CAS: 175277-99-3 | C6H4ClFS | 162.606 g/mol

$125.86 - $513.70

Chemical Identifiers

CAS 175277-99-3
Molecular Formula C6H4ClFS
Molecular Weight (g/mol) 162.606
MDL Number MFCD00052936
InChI Key KUAPPJSILOMQPC-UHFFFAOYSA-N
Synonym 2-chloro-4-fluorothiophenol, 2-chloro-4-fluorobenzene-1-thiol, benzenethiol,2-chloro-4-fluoro, 2-chloro-4-fluoro-benzenethiol, benzenethiol, 2-chloro-4-fluoro, pubchem2850, acmc-1caad, 2-chloro-4-fluoro-thiophenol, 4-fluoro-2-chloro-benzenethiol, #
PubChem CID 519446
IUPAC Name 2-chloro-4-fluorobenzenethiol
SMILES C1=CC(=C(C=C1F)Cl)S
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1349803
View Documents
Thermo Scientific Chemicals
A1349803
1 g
Each for $125.86
Only null left
 
AAA1349806
View Documents
Thermo Scientific Chemicals
A1349806
5 g
Each for $513.70
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

175277-99-3
162.606
KUAPPJSILOMQPC-UHFFFAOYSA-N
519446
C1=CC(=C(C=C1F)Cl)S
C6H4ClFS
MFCD00052936
2-chloro-4-fluorothiophenol, 2-chloro-4-fluorobenzene-1-thiol, benzenethiol,2-chloro-4-fluoro, 2-chloro-4-fluoro-benzenethiol, benzenethiol, 2-chloro-4-fluoro, pubchem2850, acmc-1caad, 2-chloro-4-fluoro-thiophenol, 4-fluoro-2-chloro-benzenethiol, #
2-chloro-4-fluorobenzenethiol

Specifications

175277-99-3
C6H4ClFS
MFCD00052936
8478226
2-chloro-4-fluorothiophenol, 2-chloro-4-fluorobenzene-1-thiol, benzenethiol,2-chloro-4-fluoro, 2-chloro-4-fluoro-benzenethiol, benzenethiol, 2-chloro-4-fluoro, pubchem2850, acmc-1caad, 2-chloro-4-fluoro-thiophenol, 4-fluoro-2-chloro-benzenethiol, #
C1=CC(=C(C=C1F)Cl)S
162.606
162.61
2-Chloro-4-fluorothiophenol
86°C to 88°C (15 mmHg)
1.567
1 g
Air Sensitive
KUAPPJSILOMQPC-UHFFFAOYSA-N
2-chloro-4-fluorobenzenethiol
519446
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H332-H315-H319-H335
Toxic if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only