missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-4-iodo-3-methylpyridine, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H5010706

 View more versions of this product

Catalog No. AAH5010706

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Chloro-4-iodo-3-methylpyridine
106°C to 107°C
MFCD03095293
Light sensitive
WHCRABJNJRAJIP-UHFFFAOYSA-N
2-chloro-4-iodo-3-methylpyridine
10083628
95%
153034-88-9
C6H5ClIN
5 g
2-chloro-4-iodo-3-picoline, 2-chloro-3-methyl-4-iodopyridine, 2-chloro-4-iodo-3-methyl-pyridine, pyridine, 2-chloro-4-iodo-3-methyl, 2-chloro-4-iodo-3-methylpyridne, pubchem17059, 2-chloranyl-4-iodanyl-3-methyl-pyridine, 2-chloro-4-iodo-3-methylpyridine, 2-chloro-4-iodo-3-methyl-pyridine; 2-chloro-4-iodo-3-picoline
CC1=C(C=CN=C1Cl)I
253.467
253.47

Chemical Identifiers

153034-88-9
253.467
WHCRABJNJRAJIP-UHFFFAOYSA-N
10083628
CC1=C(C=CN=C1Cl)I
C6H5ClIN
MFCD03095293
2-chloro-4-iodo-3-picoline, 2-chloro-3-methyl-4-iodopyridine, 2-chloro-4-iodo-3-methyl-pyridine, pyridine, 2-chloro-4-iodo-3-methyl, 2-chloro-4-iodo-3-methylpyridne, pubchem17059, 2-chloranyl-4-iodanyl-3-methyl-pyridine, 2-chloro-4-iodo-3-methylpyridine, 2-chloro-4-iodo-3-methyl-pyridine; 2-chloro-4-iodo-3-picoline
2-chloro-4-iodo-3-methylpyridine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only