missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-4-methoxytoluene 95.0+%, TCI America™
$192.00 - $598.37
Chemical Identifiers
| CAS | 54788-38-4 |
|---|---|
| Molecular Formula | C8H9ClO |
| Molecular Weight (g/mol) | 156.609 |
| MDL Number | MFCD00087376 |
| InChI Key | ZMZVVVASCILFJL-UHFFFAOYSA-N |
| Synonym | 3-chloro-4-methylanisole, 2-chloro-4-methoxytoluene, 2-chloro-4-methoxy-1-methyl-benzene, benzene, 2-chloro-4-methoxy-1-methyl, pubchem4101, acmc-209xb6, ksc602k7p, benzene,2-chloro-4-methoxy-1-methyl |
| PubChem CID | 6951757 |
| IUPAC Name | 2-chloro-4-methoxy-1-methylbenzene |
| SMILES | CC1=C(C=C(C=C1)OC)Cl |
Chemical Identifiers
| 54788-38-4 | |
| 156.609 | |
| ZMZVVVASCILFJL-UHFFFAOYSA-N | |
| 6951757 | |
| CC1=C(C=C(C=C1)OC)Cl |
| C8H9ClO | |
| MFCD00087376 | |
| 3-chloro-4-methylanisole, 2-chloro-4-methoxytoluene, 2-chloro-4-methoxy-1-methyl-benzene, benzene, 2-chloro-4-methoxy-1-methyl, pubchem4101, acmc-209xb6, ksc602k7p, benzene,2-chloro-4-methoxy-1-methyl | |
| 2-chloro-4-methoxy-1-methylbenzene |
Specifications
| 54788-38-4 | |
| 213°C | |
| MFCD00087376 | |
| 3-chloro-4-methylanisole, 2-chloro-4-methoxytoluene, 2-chloro-4-methoxy-1-methyl-benzene, benzene, 2-chloro-4-methoxy-1-methyl, pubchem4101, acmc-209xb6, ksc602k7p, benzene,2-chloro-4-methoxy-1-methyl | |
| CC1=C(C=C(C=C1)OC)Cl | |
| 156.609 | |
| 156.61 | |
| Liquid |
| Yellow | |
| C8H9ClO | |
| 5 g | |
| ZMZVVVASCILFJL-UHFFFAOYSA-N | |
| 2-chloro-4-methoxy-1-methylbenzene | |
| 6951757 | |
| ≥95.0% (GC) | |
| 2-Chloro-4-methoxytoluene |
Safety and Handling
TSCA : No