missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-chloro-4-methylaniline, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 154670250

 View more versions of this product

Catalog No. AC154670250



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Chloro-4-methylaniline
97.5
7°C
1.1510g/mL
99°C
97.5% min. (GC)
C7H8ClN
ClC6H3(CH3)NH2
25 mL
1.151
XGYLSRFSXKAYCR-UHFFFAOYSA-N
2-chloro-4-methylaniline
12007
98%
615-65-6
100.0
Brown to Yellow
223°C to 225°C
Authentic
Glass bottle
1.5740 to 1.576
MFCD00007666
12, 989
2-chloro-p-toluidine, benzenamine, 2-chloro-4-methyl, 4-amino-3-chlorotoluene, p-toluidine, 2-chloro, 2-chlor-4-toluidin, 4-methyl-2-chloroaniline, unii-k59isj2ko1, 2-chlor-4-toluidin czech, ccris 2887, 3-chloro-4-aminotoluene
CC1=CC(=C(C=C1)N)Cl
141.6
141.6
Liquid

Chemical Identifiers

615-65-6
141.6
XGYLSRFSXKAYCR-UHFFFAOYSA-N
12007
CC1=CC(=C(C=C1)N)Cl
C7H8ClN
MFCD00007666
2-chloro-p-toluidine, benzenamine, 2-chloro-4-methyl, 4-amino-3-chlorotoluene, p-toluidine, 2-chloro, 2-chlor-4-toluidin, 4-methyl-2-chloroaniline, unii-k59isj2ko1, 2-chlor-4-toluidin czech, ccris 2887, 3-chloro-4-aminotoluene
2-chloro-4-methylaniline
Safety and Handling

Safety and Handling

GHS H Statement
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye damage.
Causes skin irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin t

GHS Signal Word: Danger

EINECSNumber : 210-440-7

Recommended Storage : Product may darken during storage

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only