missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-4-(trifluoromethyl)anisole, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H3355403

 View more versions of this product

Catalog No. AAH3355403


Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
Specifications

Specifications

2-Chloro-4-(trifluoromethyl)anisole
C8H6ClF3O
MFCD13185462
3-chloro-4-methoxybenzotrifluoride, 2-chloro-1-methoxy-4-trifluoromethyl benzene, 2-chloro-4-trifluoromethyl anisole, 2-methoxy-5-trifluoromethyl chlorobenzene, 2-chloro-4-trifluoromethyl phenyl methyl ether
VVRXOIBFPBBYRU-UHFFFAOYSA-N
2-chloro-1-methoxy-4-(trifluoromethyl)benzene
54413375
97%
1214340-18-7
1.4755
1 g
Sparingly soluble in water.(0.26g/L) (25°C),
COC1=C(Cl)C=C(C=C1)C(F)(F)F
210.58
210.58

Chemical Identifiers

1214340-18-7
210.58
VVRXOIBFPBBYRU-UHFFFAOYSA-N
54413375
COC1=C(Cl)C=C(C=C1)C(F)(F)F
C8H6ClF3O
MFCD13185462
3-chloro-4-methoxybenzotrifluoride, 2-chloro-1-methoxy-4-trifluoromethyl benzene, 2-chloro-4-trifluoromethyl anisole, 2-methoxy-5-trifluoromethyl chlorobenzene, 2-chloro-4-trifluoromethyl phenyl methyl ether
2-chloro-1-methoxy-4-(trifluoromethyl)benzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions