missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 98.0+%, TCI America™

Supplier:  TCI America C31365G

 View more versions of this product

Catalog No. C31365G


Explore available promotions

Description

Description

Specifications

Specifications

2-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
87°C
C11H15BClNO2
5 g
IEFBTHROULVUAH-UHFFFAOYSA-N
2-chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
2794719
≥98.0% (GC,T)
444120-94-9
White
MFCD03412793
2-chloro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-chloropyridine-5-boronic acid pinacol ester, 2-chloro-5-pyridylboronic acid pinacol ester, 6-chloropyridine-3-boronic acid pinacol ester, 2-chloropyridine-5-boronicacidpinacolester, 2-chloro-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine, 6-chloro-3-pyridineboronic acid pinacol ester, 6-chloropyridin-3-ylboronic acid pinacol ester, 2-chloro-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-6-chloro-3-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)Cl
239.506
239.51
Crystalline Powder

Chemical Identifiers

444120-94-9
239.506
IEFBTHROULVUAH-UHFFFAOYSA-N
2794719
B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)Cl
C11H15BClNO2
MFCD03412793
2-chloro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-chloropyridine-5-boronic acid pinacol ester, 2-chloro-5-pyridylboronic acid pinacol ester, 6-chloropyridine-3-boronic acid pinacol ester, 2-chloropyridine-5-boronicacidpinacolester, 2-chloro-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine, 6-chloro-3-pyridineboronic acid pinacol ester, 6-chloropyridin-3-ylboronic acid pinacol ester, 2-chloro-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-6-chloro-3-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
2-chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions