Learn More
2-Chloro-5-fluoropyrimidine, 97%, Thermo Scientific Chemicals
$298.58 - $1133.99
Chemical Identifiers
CAS | 62802-42-0 |
---|---|
Molecular Formula | C4H2ClFN2 |
Molecular Weight (g/mol) | 132.522 |
MDL Number | MFCD03788197 |
InChI Key | AGYUQBNABXVWMS-UHFFFAOYSA-N |
Synonym | 2-chloro-5-fluoro-pyrimidine, 5-fluoro-2-chloropyrimidine, pyrimidine, 2-chloro-5-fluoro, 2-chlor-5-fluorpyrimidin, 2-chloro-5-fluoro pyrimidine, pubchem4684, acmc-1b6zl, ksc352s4h, #, 2-chloro-5-fluoro-1,3-diazine |
PubChem CID | 583466 |
IUPAC Name | 2-chloro-5-fluoropyrimidine |
SMILES | C1=C(C=NC(=N1)Cl)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5505503
|
Thermo Scientific Chemicals
H5505503 |
1 g |
Each for $298.58
|
|
|||||
AAH5505506
|
Thermo Scientific Chemicals
H5505506 |
5 g |
Each for $1,133.99
|
|
|||||
Description
2-Chloro-5-fluoropyrimidine is used to prepare 2,4-disubstituted-5-fluoropyrimidine , which is a biologically active molecule seen in anticancer agents like 5-fluorouracil.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Chloro-5-fluoropyrimidine is used to prepare 2,4-disubstituted-5-fluoropyrimidine , which is a biologically active molecule seen in anticancer agents like 5-fluorouracil.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
62802-42-0 | |
132.522 | |
AGYUQBNABXVWMS-UHFFFAOYSA-N | |
583466 | |
C1=C(C=NC(=N1)Cl)F |
C4H2ClFN2 | |
MFCD03788197 | |
2-chloro-5-fluoro-pyrimidine, 5-fluoro-2-chloropyrimidine, pyrimidine, 2-chloro-5-fluoro, 2-chlor-5-fluorpyrimidin, 2-chloro-5-fluoro pyrimidine, pubchem4684, acmc-1b6zl, ksc352s4h, #, 2-chloro-5-fluoro-1,3-diazine | |
2-chloro-5-fluoropyrimidine |
Specifications
62802-42-0 | |
65°C (149°F) | |
1.503 | |
1 g | |
2-chloro-5-fluoro-pyrimidine, 5-fluoro-2-chloropyrimidine, pyrimidine, 2-chloro-5-fluoro, 2-chlor-5-fluorpyrimidin, 2-chloro-5-fluoro pyrimidine, pubchem4684, acmc-1b6zl, ksc352s4h, #, 2-chloro-5-fluoro-1,3-diazine | |
C1=C(C=NC(=N1)Cl)F | |
132.522 | |
132.52 | |
2-Chloro-5-fluoropyrimidine |
1.073 | |
C4H2ClFN2 | |
MFCD03788197 | |
UN3267 | |
AGYUQBNABXVWMS-UHFFFAOYSA-N | |
2-chloro-5-fluoropyrimidine | |
583466 | |
97% |
Safety and Handling
GHS H Statement
H301-H314-H318-H227
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H227-H302+H332-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only