missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-6-fluorobenzoyl Chloride 98.0+%, TCI America™
$33.56 - $134.21
Chemical Identifiers
| CAS | 79455-63-3 |
|---|---|
| Molecular Formula | C7H3Cl2FO |
| Molecular Weight (g/mol) | 192.998 |
| MDL Number | MFCD00178746 |
| InChI Key | GFNAJZAKJGKJCS-UHFFFAOYSA-N |
| Synonym | 2-chloro-6-fluorobenzoylchloride, 2-chloro-6-fluorobenzene-1-carbonyl chloride, 2-chloro-6-fluoro-benzoyl chloride, 6-chloro-2-fluorobenzoyl chloride, benzoyl chloride, 2-chloro-6-fluoro, acmc-1behg, 2-fluoro-6-chlorobenzoyl chloride, attercop-chm at108371, 2-chloro-6-fluoro benzoyl chloride, 2-chloranyl-6-fluoranyl-benzoyl chloride |
| PubChem CID | 2736547 |
| IUPAC Name | 2-chloro-6-fluorobenzoyl chloride |
| SMILES | C1=CC(=C(C(=C1)Cl)C(=O)Cl)F |
Chemical Identifiers
| 79455-63-3 | |
| 192.998 | |
| GFNAJZAKJGKJCS-UHFFFAOYSA-N | |
| 2736547 | |
| C1=CC(=C(C(=C1)Cl)C(=O)Cl)F |
| C7H3Cl2FO | |
| MFCD00178746 | |
| 2-chloro-6-fluorobenzoylchloride, 2-chloro-6-fluorobenzene-1-carbonyl chloride, 2-chloro-6-fluoro-benzoyl chloride, 6-chloro-2-fluorobenzoyl chloride, benzoyl chloride, 2-chloro-6-fluoro, acmc-1behg, 2-fluoro-6-chlorobenzoyl chloride, attercop-chm at108371, 2-chloro-6-fluoro benzoyl chloride, 2-chloranyl-6-fluoranyl-benzoyl chloride | |
| 2-chloro-6-fluorobenzoyl chloride |
Specifications
| 79455-63-3 | |
| C7H3Cl2FO | |
| 5 g | |
| 2-chloro-6-fluorobenzoylchloride, 2-chloro-6-fluorobenzene-1-carbonyl chloride, 2-chloro-6-fluoro-benzoyl chloride, 6-chloro-2-fluorobenzoyl chloride, benzoyl chloride, 2-chloro-6-fluoro, acmc-1behg, 2-fluoro-6-chlorobenzoyl chloride, attercop-chm at108371, 2-chloro-6-fluoro benzoyl chloride, 2-chloranyl-6-fluoranyl-benzoyl chloride | |
| C1=CC(=C(C(=C1)Cl)C(=O)Cl)F | |
| 192.998 | |
| 193.00 | |
| Liquid |
| Yellow | |
| MFCD00178746 | |
| 3265 | |
| GFNAJZAKJGKJCS-UHFFFAOYSA-N | |
| 2-chloro-6-fluorobenzoyl chloride | |
| 2736547 | |
| ≥98.0% (GC,T) | |
| 2-Chloro-6-fluorobenzoyl Chloride |
Safety and Handling
TSCA : No