missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Chloro-6-fluorophenylacetic acid, 98%, Thermo Scientific Chemicals
$217.32 - $217.32
Chemical Identifiers
CAS | 37777-76-7 |
---|---|
Molecular Formula | C8H6ClFO2 |
Molecular Weight (g/mol) | 188.582 |
MDL Number | MFCD00004319 |
InChI Key | GUAIAAXDEJZRBP-UHFFFAOYSA-N |
Synonym | 2-chloro-6-fluorophenylacetic acid, 2-2-chloro-6-fluorophenyl acetic acid, 2-chloro-6-fluorophenyl acetic acid, 2-chloro-6-fluorophenylaceticacid, benzeneacetic acid, 2-chloro-6-fluoro, 2-chloro-6-fluoro-phenyl-acetic acid, pubchem4254, acmc-1aewm, rarechem al bo 0114 |
PubChem CID | 123464 |
IUPAC Name | 2-(2-chloro-6-fluorophenyl)acetic acid |
SMILES | C1=CC(=C(C(=C1)Cl)CC(=O)O)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1046014
|
Thermo Scientific Chemicals
A1046014 |
25 g |
Each for $217.32
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
37777-76-7 | |
188.582 | |
GUAIAAXDEJZRBP-UHFFFAOYSA-N | |
123464 | |
C1=CC(=C(C(=C1)Cl)CC(=O)O)F |
C8H6ClFO2 | |
MFCD00004319 | |
2-chloro-6-fluorophenylacetic acid, 2-2-chloro-6-fluorophenyl acetic acid, 2-chloro-6-fluorophenyl acetic acid, 2-chloro-6-fluorophenylaceticacid, benzeneacetic acid, 2-chloro-6-fluoro, 2-chloro-6-fluoro-phenyl-acetic acid, pubchem4254, acmc-1aewm, rarechem al bo 0114 | |
2-(2-chloro-6-fluorophenyl)acetic acid |
Specifications
37777-76-7 | |
C8H6ClFO2 | |
25 g | |
2-chloro-6-fluorophenylacetic acid, 2-2-chloro-6-fluorophenyl acetic acid, 2-chloro-6-fluorophenyl acetic acid, 2-chloro-6-fluorophenylaceticacid, benzeneacetic acid, 2-chloro-6-fluoro, 2-chloro-6-fluoro-phenyl-acetic acid, pubchem4254, acmc-1aewm, rarechem al bo 0114 | |
C1=CC(=C(C(=C1)Cl)CC(=O)O)F | |
188.582 | |
188.59 | |
2-Chloro-6-fluorophenylacetic acid |
120°C to 124°C | |
MFCD00004319 | |
4401142 | |
GUAIAAXDEJZRBP-UHFFFAOYSA-N | |
2-(2-chloro-6-fluorophenyl)acetic acid | |
123464 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 253-661-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only