missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-6-methylaniline 98.0+%, TCI America™

Supplier:  TCI America A19595G

 View more versions of this product

Catalog No. A19595G


Explore available promotions

Description

Description

Specifications

Specifications

2-Chloro-6-methylaniline
12°C
C7H8ClN
5 g
6-chloro-2-methylaniline, 6-chloro-o-toluidine, 2-amino-3-chlorotoluene, benzenamine, 2-chloro-6-methyl, 3-chloro-2-aminotoluene, 6-chloro-2-toluidine, 2-methyl-6-chloroaniline, o-toluidine, 6-chloro, 2-chloro-6-methyl-aniline, 2-chloro-6-methylaninile
CC1=CC=CC(Cl)=C1N
141.60
141.60
Liquid
87-63-8
215°C
MFCD00007679
3429
WFNLHDJJZSJARK-UHFFFAOYSA-N
2-chloro-6-methylaniline
6897
≥98.0% (GC,T)

Chemical Identifiers

87-63-8
141.60
WFNLHDJJZSJARK-UHFFFAOYSA-N
6897
CC1=CC=CC(Cl)=C1N
C7H8ClN
MFCD00007679
6-chloro-2-methylaniline, 6-chloro-o-toluidine, 2-amino-3-chlorotoluene, benzenamine, 2-chloro-6-methyl, 3-chloro-2-aminotoluene, 6-chloro-2-toluidine, 2-methyl-6-chloroaniline, o-toluidine, 6-chloro, 2-chloro-6-methyl-aniline, 2-chloro-6-methylaninile
2-chloro-6-methylaniline
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (9)-0073&(3)-0265

RTECSNumber : XU5100000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions