missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-6-methylquinoline-3-carboxaldehyde 96.0+%, TCI America™
Supplier: TCI America C29535G
Specifications
| 2-Chloro-6-methylquinoline-3-carboxaldehyde | |
| 124°C | |
| C11H8ClNO | |
| 5 g | |
| FSLNYYZJXMGKHK-UHFFFAOYSA-N | |
| 2-chloro-6-methylquinoline-3-carbaldehyde | |
| 689081 | |
| ≥96.0% (GC) |
| 73568-27-1 | |
| Yellow | |
| MFCD01912773 | |
| 2-Chloro-3-formyl-6-methylquinoline | |
| CC1=CC=C2N=C(Cl)C(C=O)=CC2=C1 | |
| 205.64 | |
| 205.64 | |
| Crystalline Powder |
Chemical Identifiers
| 73568-27-1 | |
| 205.64 | |
| FSLNYYZJXMGKHK-UHFFFAOYSA-N | |
| 689081 | |
| CC1=CC=C2N=C(Cl)C(C=O)=CC2=C1 |
| C11H8ClNO | |
| MFCD01912773 | |
| 2-Chloro-3-formyl-6-methylquinoline | |
| 2-chloro-6-methylquinoline-3-carbaldehyde |
Safety and Handling
TSCA : No