Learn More
2-Chloroacetamide, 98%, Thermo Scientific Chemicals
$62.32 - $88.79
Chemical Identifiers
CAS | 79-07-2 |
---|---|
Molecular Formula | C2H4ClNO |
Molecular Weight (g/mol) | 93.51 |
MDL Number | MFCD00008027 |
InChI Key | VXIVSQZSERGHQP-UHFFFAOYSA-N |
Synonym | chloroacetamide, acetamide, 2-chloro, chloracetamide, 2-chloroethanamide, microcide, mergal af, chloracetamid, usaf do-29, 2-chloracetamide, 2-chloro-acetamide |
PubChem CID | 6580 |
IUPAC Name | 2-chloroacetamide |
SMILES | NC(=O)CCl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC148410050
|
Thermo Scientific Chemicals
148410050 |
5 g | Glass bottle |
Each for $62.32
|
|
||||
AC148415000
|
Thermo Scientific Chemicals
148415000 |
500 g | Plastic bottle |
Each for $88.79
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
79-07-2 | |
93.51 | |
VXIVSQZSERGHQP-UHFFFAOYSA-N | |
6580 | |
NC(=O)CCl |
C2H4ClNO | |
MFCD00008027 | |
chloroacetamide, acetamide, 2-chloro, chloracetamide, 2-chloroethanamide, microcide, mergal af, chloracetamid, usaf do-29, 2-chloracetamide, 2-chloro-acetamide | |
2-chloroacetamide |
Specifications
79-07-2 | |
100.0 | |
White | |
224.5°C | |
Authentic | |
Glass bottle | |
ClCH2CONH2 | |
5 g | |
13,70 | |
15, 2110 | |
Solubility in water: 90g/L (20°C). Other solubilities: soluble in alcohol, insoluble in ether | |
NC(=O)CCl | |
93.51 | |
93.51 | |
Crystalline Powder |
97.5 | |
116.0°C to 120.0°C | |
0.8400g/mL | |
170°C | |
97.5% min. (GC) | |
C2H4ClNO | |
MFCD00008027 | |
02, 199 | |
0.84 | |
chloroacetamide, acetamide, 2-chloro, chloracetamide, 2-chloroethanamide, microcide, mergal af, chloracetamid, usaf do-29, 2-chloracetamide, 2-chloro-acetamide | |
VXIVSQZSERGHQP-UHFFFAOYSA-N | |
2-chloroacetamide | |
6580 | |
98% | |
2-Chloroacetamide, 98% |
Safety and Handling
GHS H Statement:
Toxic if swallowed.
May cause an allergic skin reaction.
Suspected of damaging fertility.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Use personal protective equipment as required.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 201-174-2
RTECSNumber : AB5075000
TSCA : TSCA
RUO – Research Use Only