Learn More
2-Chloroaniline hydrochloride, 97%
CAS: 137-04-2 | C6H7Cl2N | 164.03 g/mol
$77.51 - $204.78
Chemical Identifiers
| CAS | 137-04-2 |
|---|---|
| Molecular Formula | C6H7Cl2N |
| Molecular Weight (g/mol) | 164.03 |
| MDL Number | MFCD00053282 |
| InChI Key | DRGIDRZFKRLQTE-UHFFFAOYSA-N |
| Synonym | 2-chloroaniline hydrochloride, 2-chloroanilinium chloride, benzenamine, 2-chloro-, hydrochloride, benzenamine, 2-chloro-, hydrochloride 1:1, 2-chlorophenylamine, chloride, acmc-209cai, o-chloroaniline hydrochloride, 95-51-2 parent, 2-chloranylaniline hydrochloride, benzenamine,2-chloro-,hydrochloride 1:1 |
| PubChem CID | 67298 |
| IUPAC Name | 2-chloroaniline;hydrochloride |
| SMILES | [H+].[Cl-].NC1=CC=CC=C1Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1348314
|
Thermo Scientific Chemicals
L1348314 |
25 g |
N/A
|
|
|||||
|
AAL1348322
|
Thermo Scientific Chemicals
L1348322 |
100 g |
N/A
|
|
|||||
Description
2-Chloroaniline hydrochloride, is used as ice dye basement, used for towels, colored yarn and cloth of cotton, viscose fabric dyeing and printing of chromogenic agent. It is also used as an intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Chloroaniline hydrochloride, is used as ice dye basement, used for towels, colored yarn and cloth of cotton, viscose fabric dyeing and printing of chromogenic agent. It is also used as an intermediate in organic synthesis.
Solubility
It is soluble in water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Chemical Identifiers
| 137-04-2 | |
| 164.03 | |
| DRGIDRZFKRLQTE-UHFFFAOYSA-N | |
| 67298 | |
| [H+].[Cl-].NC1=CC=CC=C1Cl |
| C6H7Cl2N | |
| MFCD00053282 | |
| 2-chloroaniline hydrochloride, 2-chloroanilinium chloride, benzenamine, 2-chloro-, hydrochloride, benzenamine, 2-chloro-, hydrochloride 1:1, 2-chlorophenylamine, chloride, acmc-209cai, o-chloroaniline hydrochloride, 95-51-2 parent, 2-chloranylaniline hydrochloride, benzenamine,2-chloro-,hydrochloride 1:1 | |
| 2-chloroaniline;hydrochloride |
Specifications
| 137-04-2 | |
| Characteristic | |
| MFCD00053282 | |
| UN2018 | |
| 2-chloroaniline hydrochloride, 2-chloroanilinium chloride, benzenamine, 2-chloro-, hydrochloride, benzenamine, 2-chloro-, hydrochloride 1:1, 2-chlorophenylamine, chloride, acmc-209cai, o-chloroaniline hydrochloride, 95-51-2 parent, 2-chloranylaniline hydrochloride, benzenamine,2-chloro-,hydrochloride 1:1 | |
| DRGIDRZFKRLQTE-UHFFFAOYSA-N | |
| 2-chloroaniline;hydrochloride | |
| 67298 | |
| 97% |
| ∼220°C (sublimation) | |
| C6H7Cl2N | |
| 25 g | |
| 3624280 | |
| It is soluble in water. | |
| [H+].[Cl-].NC1=CC=CC=C1Cl | |
| 164.03 | |
| 164.03 | |
| 2-Chloroaniline hydrochloride |
Safety and Handling
GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P501c
H301+H311+H331-H373
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROANILINES, SOLID
EINECSNumber : 205-274-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only