Learn More
2-Chlorobenzoic acid, 98%, Thermo Scientific Chemicals
$30.85 - $86.91
Chemical Identifiers
CAS | 118-91-2 |
---|---|
Molecular Formula | C7H5ClO2 |
Molecular Weight (g/mol) | 156.57 |
MDL Number | MFCD00002412 |
InChI Key | IKCLCGXPQILATA-UHFFFAOYSA-N |
Synonym | o-chlorobenzoic acid, benzoic acid, o-chloro, benzoic acid, 2-chloro, 2-cba, kyselina o-chlorbenzoova, chlorobenzoic acid, ocba, benzoic acid, chloro, 2-chloro-benzoic acid, chlorobenzoate |
PubChem CID | 8374 |
ChEBI | CHEBI:30793 |
IUPAC Name | 2-chlorobenzoic acid |
SMILES | C1=CC=C(C(=C1)C(=O)O)Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC159241000
|
Thermo Scientific Chemicals
159241000 |
100 g | Plastic bottle |
Each for $30.85
|
|
||||
AC159245000
|
Thermo Scientific Chemicals
159245000 |
500 g | Plastic bottle |
Each for $86.91
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
118-91-2 | |
156.57 | |
IKCLCGXPQILATA-UHFFFAOYSA-N | |
8374 | |
2-chlorobenzoic acid |
C7H5ClO2 | |
MFCD00002412 | |
o-chlorobenzoic acid, benzoic acid, o-chloro, benzoic acid, 2-chloro, 2-cba, kyselina o-chlorbenzoova, chlorobenzoic acid, ocba, benzoic acid, chloro, 2-chloro-benzoic acid, chlorobenzoate | |
CHEBI:30793 | |
C1=CC=C(C(=C1)C(=O)O)Cl |
Specifications
118-91-2 | |
100.0 | |
White | |
173°C | |
97.5% min. (HPLC) | |
C7H5ClO2 | |
MFCD00002412 | |
09, 334 | |
o-chlorobenzoic acid, benzoic acid, o-chloro, benzoic acid, 2-chloro, 2-cba, kyselina o-chlorbenzoova, chlorobenzoic acid, ocba, benzoic acid, chloro, 2-chloro-benzoic acid, chlorobenzoate | |
IKCLCGXPQILATA-UHFFFAOYSA-N | |
2-chlorobenzoic acid | |
8374 | |
156.57 | |
Powder |
97.5 | |
139.0°C to 143.0°C | |
285.0°C | |
Authentic | |
Plastic bottle | |
ClC6H4CO2H | |
100 g | |
15, 2125 | |
Solubility in water: soluble in hot water. Other solubilities: freely soluble in alcohol and ether | |
C1=CC=C(C(=C1)C(=O)O)Cl | |
156.57 | |
CHEBI:30793 | |
98% | |
2-Chlorobenzoic acid, 98% |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clo
GHS Signal Word: Warning
EINECSNumber : 204-285-4
RTECSNumber : DG4976000
TSCA : TSCA
RUO – Research Use Only