missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzothiazole 98.0+%, TCI America™

$104.38 - $590.90

Chemical Identifiers

CAS 615-20-3
Molecular Formula C7H4ClNS
Molecular Weight (g/mol) 169.626
MDL Number MFCD00005776
InChI Key BSQLQMLFTHJVKS-UHFFFAOYSA-N
Synonym 2-chlorobenzothiazole, benzothiazole, 2-chloro, 2-chlorobenzo d thiazole, 2-benzothiazolyl chloride, usaf ek-2784, 2-chloro-benzothiazole, benzothiazole, chloro, unii-fn1t1y0p5p, fn1t1y0p5p, 2-chlorobenzthiazole
PubChem CID 11987
IUPAC Name 2-chloro-1,3-benzothiazole
SMILES C1=CC=C2C(=C1)N=C(S2)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C033225G
View Documents
TCI America
C033225G
25 g
Each for $104.38
Only null left
 
C0332250G
View Documents
TCI America
C0332250G
250 g
Each for $590.90
Only null left
 
Description

Description

Specifications

Chemical Identifiers

615-20-3
169.626
BSQLQMLFTHJVKS-UHFFFAOYSA-N
11987
C1=CC=C2C(=C1)N=C(S2)Cl
C7H4ClNS
MFCD00005776
2-chlorobenzothiazole, benzothiazole, 2-chloro, 2-chlorobenzo d thiazole, 2-benzothiazolyl chloride, usaf ek-2784, 2-chloro-benzothiazole, benzothiazole, chloro, unii-fn1t1y0p5p, fn1t1y0p5p, 2-chlorobenzthiazole
2-chloro-1,3-benzothiazole

Specifications

615-20-3
248°C
MFCD00005776
2-chlorobenzothiazole, benzothiazole, 2-chloro, 2-chlorobenzo d thiazole, 2-benzothiazolyl chloride, usaf ek-2784, 2-chloro-benzothiazole, benzothiazole, chloro, unii-fn1t1y0p5p, fn1t1y0p5p, 2-chlorobenzthiazole
C1=CC=C2C(=C1)N=C(S2)Cl
169.626
169.63
Crystalline Lumps
White-Yellow
C7H4ClNS
25 g
BSQLQMLFTHJVKS-UHFFFAOYSA-N
2-chloro-1,3-benzothiazole
11987
≥98.0% (GC)
2-Chlorobenzothiazole
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-6513

RTECSNumber : DL3150000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'