Learn More
2-Chlorobutane, 98+%, Thermo Scientific Chemicals
$54.04 - $239.23
Chemical Identifiers
CAS | 78-86-4 |
---|---|
Molecular Formula | C4H9Cl |
Molecular Weight (g/mol) | 92.566 |
MDL Number | MFCD00000871 |
InChI Key | BSPCSKHALVHRSR-UHFFFAOYSA-N |
Synonym | sec-butyl chloride, butane, 2-chloro, 1-methylpropyl chloride, 2-chloro-butane, ccris 9391, alkanes, c11-14, 2-chloro, sec.-butyl chloride, sjphadilup@, sec-c4h9cl, acmc-209pfl |
PubChem CID | 6563 |
IUPAC Name | 2-chlorobutane |
SMILES | CCC(C)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
AAB21586AD
|
Thermo Scientific Chemicals
B21586AD |
50 mL |
Each for $54.04
|
|
|||||
AAB21586AK
|
Thermo Scientific Chemicals
B21586AK |
250 mL |
Each for $89.86
|
|
|||||
AAB21586AU
|
Thermo Scientific Chemicals
B21586AU |
1000 mL |
Each for $239.23
|
|
|||||
Description
2-Chlorobutane is used as a solvent and intermediate in organic synthesis. Further, it is involved in the preparation of active pharmaceutical ingredients, plasticizer, rubber, resin and surfactants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Chlorobutane is used as a solvent and intermediate in organic synthesis. Further, it is involved in the preparation of active pharmaceutical ingredients, plasticizer, rubber, resin and surfactants.
Solubility
Immiscible with water.
Notes
Incompatible with metals, copper, strong oxidizing agents and strong bases.
Chemical Identifiers
78-86-4 | |
92.566 | |
BSPCSKHALVHRSR-UHFFFAOYSA-N | |
6563 | |
CCC(C)Cl |
C4H9Cl | |
MFCD00000871 | |
sec-butyl chloride, butane, 2-chloro, 1-methylpropyl chloride, 2-chloro-butane, ccris 9391, alkanes, c11-14, 2-chloro, sec.-butyl chloride, sjphadilup@, sec-c4h9cl, acmc-209pfl | |
2-chlorobutane |
Specifications
78-86-4 | |
0.873 | |
−15°C (5°F) | |
Sweet | |
1.397 | |
50 mL | |
1718770 | |
sec-butyl chloride, butane, 2-chloro, 1-methylpropyl chloride, 2-chloro-butane, ccris 9391, alkanes, c11-14, 2-chloro, sec.-butyl chloride, sjphadilup@, sec-c4h9cl, acmc-209pfl | |
BSPCSKHALVHRSR-UHFFFAOYSA-N | |
2-chlorobutane | |
6563 | |
≥98% | |
2-Chlorobutane |
-141°C to -140°C | |
67°C to 69°C | |
98+% | |
C4H9Cl | |
MFCD00000871 | |
UN1127 | |
14,1561 | |
Immiscible with water. | |
CCC(C)Cl | |
92.566 | |
92.57 | |
Liquid |
Safety and Handling
GHS H Statement
H225
Highly flammable liquid and vapour.
P210-P233-P234-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P390-P501c
H225-H290
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CHLOROBUTANES
EINECSNumber : 201-151-7
RTECSNumber : EJ6475000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only