Learn More
2-Chlorobutyric acid, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L2008314
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Chlorobutyric acid | |
1.19 | |
112°C (233°F) | |
1.439 | |
25 g | |
1720944 | |
RVBUZBPJAGZHSQ-UHFFFAOYSA-N | |
2-chlorobutanoic acid | |
20137 | |
97% |
4170-24-5 | |
90°C to 92°C (12 mmHg) | |
C4H7ClO2 | |
MFCD00021697 | |
UN3265 | |
2-chlorobutyric acid, butanoic acid, 2-chloro, butyric acid, 2-chloro, ccris 8992, 2-chloro-butyric acid, 2-chlorbutyric acid, acmc-209huc, acmc-209lcl, 2-chloro-n-butyric acid, butanoic acid,2-chloro | |
CCC(C(=O)O)Cl | |
122.548 | |
122.55 |
Chemical Identifiers
4170-24-5 | |
122.548 | |
RVBUZBPJAGZHSQ-UHFFFAOYSA-N | |
20137 | |
CCC(C(=O)O)Cl |
C4H7ClO2 | |
MFCD00021697 | |
2-chlorobutyric acid, butanoic acid, 2-chloro, butyric acid, 2-chloro, ccris 8992, 2-chloro-butyric acid, 2-chlorbutyric acid, acmc-209huc, acmc-209lcl, 2-chloro-n-butyric acid, butanoic acid,2-chloro | |
2-chlorobutanoic acid |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 224-029-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only