missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloroethanol, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 036636AK

 View more versions of this product

Catalog No. AA36636AK


Explore available promotions

Description

Description

2-Chloroethanol is used in the preparation of several dyes by the alkylation of aniline derivatives with chloroethanol. It is a building block in the production of pharmaceuticals and plasticizers. Also used for manufacture of thiodiglycol. It is a solvent for cellulose acetate and ethyl cellulose, textile printing dyes, in dewaxing, refining of rosin, extraction of pine lignin and the cleaning of machines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Chloroethanol
°C to 63°C
129°C
C2H5ClO
CH2CH2OH
250 mL
878139
ethylene chlorohydrin, ethanol, 2-chloro, glycol chlorohydrin, 2-chloroethyl alcohol, chloroethanol, glycol monochlorohydrin, 2-monochloroethanol, ethylchlorohydrin, 2-chlorethanol, 2-chloro-1-ethanol
SZIFAVKTNFCBPC-UHFFFAOYSA-N
2-chloroethanol
34
80.51
Liquid
107-07-3
1.202
55°C (131°F)
1.441
MFCD00002829
UN1135
14,3793
Fully miscible with water.
C(CCl)O
80.511
CHEBI:28200
99%

Chemical Identifiers

107-07-3
80.511
SZIFAVKTNFCBPC-UHFFFAOYSA-N
34
2-chloroethanol
C2H5ClO
MFCD00002829
ethylene chlorohydrin, ethanol, 2-chloro, glycol chlorohydrin, 2-chloroethyl alcohol, chloroethanol, glycol monochlorohydrin, 2-monochloroethanol, ethylchlorohydrin, 2-chlorethanol, 2-chloro-1-ethanol
CHEBI:28200
C(CCl)O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330-H226

P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P310-P330-P363-P370+P378q-P501c

H226-H300+H310+H330

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: ETHYLENE CHLOROHYDRIN

EINECSNumber : 203-459-7

RTECSNumber : KK0875000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only