missing translation for 'onlineSavingsMsg'
Learn More

2-(Chloroethyl)benzene, 99%

CAS: 622-24-2 | C8H9Cl | 140.61 g/mol

$58.78 - $563.14

Chemical Identifiers

CAS 622-24-2
Molecular Formula C8H9Cl
Molecular Weight (g/mol) 140.61
MDL Number MFCD00000977
InChI Key MNNZINNZIQVULG-UHFFFAOYSA-N
Synonym 2-chloroethyl benzene, phenethyl chloride, 2-phenylethyl chloride, benzene, 2-chloroethyl, 1-chloro-2-phenylethane, phenethylchloride, ethylchlorobenzene, 2-phenyl-1-chloroethane, 2-chloroethyl-benzene, beta-phenethyl chloride
PubChem CID 231496
IUPAC Name 2-chloroethylbenzene
SMILES ClCCC1=CC=CC=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0489109
View Documents
Thermo Scientific Chemicals
L0489109
10 g
Each for $58.78
Only null left
 
AAL0489122
View Documents
Thermo Scientific Chemicals
L0489122
100 g
Each for $178.30
Only null left
 
AAL0489136
View Documents
Thermo Scientific Chemicals
L0489136
500 g
Each for $563.14
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

622-24-2
140.61
MNNZINNZIQVULG-UHFFFAOYSA-N
231496
ClCCC1=CC=CC=C1
C8H9Cl
MFCD00000977
2-chloroethyl benzene, phenethyl chloride, 2-phenylethyl chloride, benzene, 2-chloroethyl, 1-chloro-2-phenylethane, phenethylchloride, ethylchlorobenzene, 2-phenyl-1-chloroethane, 2-chloroethyl-benzene, beta-phenethyl chloride
2-chloroethylbenzene

Specifications

622-24-2
1.07
66°C (150°F)
1.53
10 g
2-chloroethyl benzene, phenethyl chloride, 2-phenylethyl chloride, benzene, 2-chloroethyl, 1-chloro-2-phenylethane, phenethylchloride, ethylchlorobenzene, 2-phenyl-1-chloroethane, 2-chloroethyl-benzene, beta-phenethyl chloride
ClCCC1=CC=CC=C1
140.61
140.61
2-(Chloroethyl)benzene
-60°C
82°C to 84°C (16 mmHg)
C8H9Cl
MFCD00000977
385969
MNNZINNZIQVULG-UHFFFAOYSA-N
2-chloroethylbenzene
231496
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

EINECSNumber : 210-725-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only