missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloroethyl ethyl ether, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2430822

 View more versions of this product

Catalog No. AAB2430822


Explore available promotions

Description

Description

It is used as a pharmaceutical intermediate. Photocatalytic oxidation of gaseous 2-chloroethyl ethyl sulfide over TiO2 illuminated with UV light and maintained at 25 or 80°C in air was found to suffer progressive oxidation to yield ethylene, chloroethylene , ethanol, acetaldehyde, chloroacetaldehyde, diethyl disulfide, 2-chloroethyl ethyl disulfide and bis(2-chloroethyl) disulfide as the main primary intermediates, and water (H2O), carbon dioxide (CO2), sulfur dioxide (SO2), surface sulfate ions (SO42-), and hydrogen chloride (HCl) as the final products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate. Photocatalytic oxidation of gaseous 2-chloroethyl ethyl sulfide over TiO2 illuminated with UV light and maintained at 25 or 80°C in air was found to suffer progressive oxidation to yield ethylene, chloroethylene , ethanol, acetaldehyde, chloroacetaldehyde, diethyl disulfide, 2-chloroethyl ethyl disulfide and bis(2-chloroethyl) disulfide as the main primary intermediates, and water (H2O), carbon dioxide (CO2), sulfur dioxide (SO2), surface sulfate ions (SO42-), and hydrogen chloride (HCl) as the final products.

Solubility
Miscible with water.

Notes
Store at room temperature. Incompatible with oxidizing agents.
Specifications

Specifications

2-Chloroethyl ethyl ether
-60°C
107°C to 108°C
C4H9ClO
MFCD00000974
UN3271
Miscible with water.
CCOCCCl
108.57
108.57
628-34-2
0.99
6°C (43°F)
1.412
100 g
2-chloroethyl ethyl ether, 2-ethoxyethyl chloride, 2-chloroethylethylether, 2-chloroethoxyethane, 1-chloro-2-ethoxy-ethane, ether, 2-chloroethyl ethyl, beta-chloroethyl ethyl ether, ethyl beta-chloroethyl ether, ethane, 1-chloro-2-ethoxy, ccris 9095
GPTVQTPMFOLLOA-UHFFFAOYSA-N
1-chloro-2-ethoxyethane
12341
≥98%

Chemical Identifiers

628-34-2
108.57
GPTVQTPMFOLLOA-UHFFFAOYSA-N
12341
CCOCCCl
C4H9ClO
MFCD00000974
2-chloroethyl ethyl ether, 2-ethoxyethyl chloride, 2-chloroethylethylether, 2-chloroethoxyethane, 1-chloro-2-ethoxy-ethane, ether, 2-chloroethyl ethyl, beta-chloroethyl ethyl ether, ethyl beta-chloroethyl ether, ethane, 1-chloro-2-ethoxy, ccris 9095
1-chloro-2-ethoxyethane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H331-H315-H319-H335
Highly flammable liquid and vapour.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P363-P370+P378q-P501c

H225-H302+H332-H315-H318-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.

EINECSNumber : 211-037-9

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only