Learn More
2-Chloroethyl isocyanate, 97%, Thermo Scientific Chemicals
$148.58 - $492.70
Chemical Identifiers
CAS | 1943-83-5 |
---|---|
Molecular Formula | C3H4ClNO |
Molecular Weight (g/mol) | 105.521 |
MDL Number | MFCD00002043 |
InChI Key | BCMYXYHEMGPZJN-UHFFFAOYSA-N |
Synonym | 2-chloroethyl isocyanate, 2-chloroethylisocyanate, chloroethylisocyanate, ethane, 1-chloro-2-isocyanato, 2-chlorethylisokyanat, 2-chloro ethyl isocyanate, isocyanic acid, 2-chloroethyl ester, 1-chloro-2-isocyanato-ethane, 2-chlorethylisokyanat czech, .beta.-chloroethylisocyanate |
PubChem CID | 16035 |
IUPAC Name | 1-chloro-2-isocyanatoethane |
SMILES | C(CCl)N=C=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1728914
|
Thermo Scientific Chemicals
A1728914 |
25 g |
N/A
|
|
|||||
AAA1728922
|
Thermo Scientific Chemicals
A1728922 |
100 g |
N/A
|
|
|||||
Description
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.
Solubility
Hydrolyzes in water.
Notes
Store at 4°C. Incompatible with alcohols. Amines, oxidizing agents. Hygroscopic.
Chemical Identifiers
1943-83-5 | |
105.521 | |
BCMYXYHEMGPZJN-UHFFFAOYSA-N | |
16035 | |
C(CCl)N=C=O |
C3H4ClNO | |
MFCD00002043 | |
2-chloroethyl isocyanate, 2-chloroethylisocyanate, chloroethylisocyanate, ethane, 1-chloro-2-isocyanato, 2-chlorethylisokyanat, 2-chloro ethyl isocyanate, isocyanic acid, 2-chloroethyl ester, 1-chloro-2-isocyanato-ethane, 2-chlorethylisokyanat czech, .beta.-chloroethylisocyanate | |
1-chloro-2-isocyanatoethane |
Specifications
1943-83-5 | |
135°C | |
Pungent | |
1.447 | |
25 g | |
1071429 | |
2-chloroethyl isocyanate, 2-chloroethylisocyanate, chloroethylisocyanate, ethane, 1-chloro-2-isocyanato, 2-chlorethylisokyanat, 2-chloro ethyl isocyanate, isocyanic acid, 2-chloroethyl ester, 1-chloro-2-isocyanato-ethane, 2-chlorethylisokyanat czech, .beta.-chloroethylisocyanate | |
BCMYXYHEMGPZJN-UHFFFAOYSA-N | |
1-chloro-2-isocyanatoethane | |
16035 | |
97% |
1.237 | |
56°C (132°F) | |
C3H4ClNO | |
MFCD00002043 | |
UN3080 | |
Moisture sensitive | |
Hydrolyzes in water. | |
C(CCl)N=C=O | |
105.521 | |
105.52 | |
2-Chloroethyl isocyanate |
Safety and Handling
GHS H Statement
H300-H330-H334-H314-H318-H226
Fatal if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H226-H301+H311+H331-H314-H334-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ISOCYANATES, TOXIC, FLAMMABLE, N.O.S.
EINECSNumber : 217-734-4
RTECSNumber : NQ8450000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only