missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroethyl methyl ether, 98%

CAS: 627-42-9 | C3H7ClO | 94.54 g/mol

Supplier:  Thermo Scientific Chemicals B2408136

 View more versions of this product

Catalog No. AAB2408136


Only null left
Explore available promotions

Description

Description

2-Chloroethyl methyl ether was used in the synthesis of acyclic nucleosides of thieno[2,3-d] pyrimidine derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloroethyl methyl ether was used in the synthesis of acyclic nucleosides of thieno[2,3-d] pyrimidine derivatives.

Solubility
Soluble in water 60 g/L at 20°C.

Notes
Store away from oxidizing agents, bases.
Specifications

Specifications

2-Chloroethyl methyl ether
1.04
8°C (46°F)
1.409
MFCD00000972
UN3271
Soluble in water 60g/L at 20°C.
COCCCl
94.54
94.54
627-42-9
89°C to 90°C
C3H7ClO
CH3OCH2CH2Cl
500 g
2-chloroethyl methyl ether, 2-methoxyethyl chloride, ethane, 1-chloro-2-methoxy, 2-chloroethylmethylether, unii-u890tj10ym, chloro-2-methoxyethane, chloroethyl-methyl ether, 1-chloro-2-methoxy-ethane, ether, 2-chloroethyl methyl
XTIGGAHUZJWQMD-UHFFFAOYSA-N
1-chloro-2-methoxyethane
12316
98%

Chemical Identifiers

627-42-9
94.54
XTIGGAHUZJWQMD-UHFFFAOYSA-N
12316
COCCCl
C3H7ClO
MFCD00000972
2-chloroethyl methyl ether, 2-methoxyethyl chloride, ethane, 1-chloro-2-methoxy, 2-chloroethylmethylether, unii-u890tj10ym, chloro-2-methoxyethane, chloroethyl-methyl ether, 1-chloro-2-methoxy-ethane, ether, 2-chloroethyl methyl
1-chloro-2-methoxyethane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H302-H332
Highly flammable liquid and vapour.
Harmful if swallowed.
Harmful if inhaled.

P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P312-P330-P363-P370+P378q-P372-P374-P380-P501c

H225-H302

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.

EINECSNumber : 210-999-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only