missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloroethyl phenyl sulfide, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2326506

 View more versions of this product

Catalog No. AAB2326506


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Chloroethyl phenyl sulfide
1.178
104°C (220°F)
1.583
5 g
1907116
QDXIHHOPZFCEAP-UHFFFAOYSA-N
2-chloroethylsulfanylbenzene
21702
98%
5535-49-9
245°C
C8H9ClS
MFCD00013691
UN2922
2-chloroethyl phenyl sulfide, 2-chloroethyl phenyl sulfane, 2-chloroethyl sulfanyl benzene, 2-chloroethyl thio benzene, 2-chloroethylphenylsulfide, benzene, 2-chloroethyl thio, 2-phenylthio ethyl chloride, 2-phenylthioethylchloride, acmc-1axe6
ClCCSC1=CC=CC=C1
172.67
172.68

Chemical Identifiers

5535-49-9
172.67
QDXIHHOPZFCEAP-UHFFFAOYSA-N
21702
ClCCSC1=CC=CC=C1
C8H9ClS
MFCD00013691
2-chloroethyl phenyl sulfide, 2-chloroethyl phenyl sulfane, 2-chloroethyl sulfanyl benzene, 2-chloroethyl thio benzene, 2-chloroethylphenylsulfide, benzene, 2-chloroethyl thio, 2-phenylthio ethyl chloride, 2-phenylthioethylchloride, acmc-1axe6
2-chloroethylsulfanylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302+H312+H332-H314

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 226-891-8

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only