missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroethylamine hydrochloride, 98+%

CAS: 870-24-6 | C2H7Cl2N | 115.99 g/mol

$65.67 - $794.38

Chemical Identifiers

CAS 870-24-6
Molecular Formula C2H7Cl2N
Molecular Weight (g/mol) 115.99
MDL Number MFCD00012887,MFCD00137399
InChI Key ONRREFWJTRBDRA-UHFFFAOYSA-N
Synonym 2-chloroethylamine hydrochloride, 2-chloroethanamine hydrochloride, 1-amino-2-chloroethane hydrochloride, ethanamine, 2-chloro-, hydrochloride, 2-aminoethyl chloride hydrochloride, ethanamine, 2-chloro-, hydrochloride 1:1, 2-chloroethan-1-amine hydrochloride, chlorethamine hydrochloride, 2-chloro-ethylamine hydrochloride, chlorethamine hcl
PubChem CID 9793737
IUPAC Name 2-chloroethanamine;hydrochloride
SMILES [Cl-].[NH3+]CCCl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1445518
View Documents
Thermo Scientific Chemicals
A1445518
50 g N/A
Only null left
 
AAA1445530
View Documents
Thermo Scientific Chemicals
A1445530
250 g N/A
Only null left
 
AAA144550B
View Documents
Thermo Scientific Chemicals
A144550B
1000 g N/A
Only null left
 
Description

Description

2-chloroethylamine hydrochloride is used as an intermediate in active pharmaceutical ingredients and dyes. It is also used in organic synthesis. Further, it is employed as a derivatizing reagent for amino acids, dipeptides and nucleotides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-chloroethylamine hydrochloride is used as an intermediate in active pharmaceutical ingredients and dyes. It is also used in organic synthesis. Further, it is employed as a derivatizing reagent for amino acids, dipeptides and nucleotides.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

870-24-6
115.99
ONRREFWJTRBDRA-UHFFFAOYSA-N
9793737
[Cl-].[NH3+]CCCl
C2H7Cl2N
MFCD00012887,MFCD00137399
2-chloroethylamine hydrochloride, 2-chloroethanamine hydrochloride, 1-amino-2-chloroethane hydrochloride, ethanamine, 2-chloro-, hydrochloride, 2-aminoethyl chloride hydrochloride, ethanamine, 2-chloro-, hydrochloride 1:1, 2-chloroethan-1-amine hydrochloride, chlorethamine hydrochloride, 2-chloro-ethylamine hydrochloride, chlorethamine hcl
2-chloroethanamine;hydrochloride

Specifications

870-24-6
2 to 3
C2H7Cl2N
50 g
3590304
2-chloroethylamine hydrochloride, 2-chloroethanamine hydrochloride, 1-amino-2-chloroethane hydrochloride, ethanamine, 2-chloro-, hydrochloride, 2-aminoethyl chloride hydrochloride, ethanamine, 2-chloro-, hydrochloride 1:1, 2-chloroethan-1-amine hydrochloride, chlorethamine hydrochloride, 2-chloro-ethylamine hydrochloride, chlorethamine hcl
ONRREFWJTRBDRA-UHFFFAOYSA-N
2-chloroethanamine;hydrochloride
9793737
≥98%
143°C to 148°C
Odorless
MFCD00012887,MFCD00137399
UN3261
Hygroscopic
Soluble in water.
[Cl-].[NH3+]CCCl
115.99
115.99
2-Chloroethylamine hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H341
Causes severe skin burns and eye damage.
Suspected of causing genetic defects.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 212-793-2

RTECSNumber : KR1800000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only