Learn More
2-Chloromandelic acid, 98%, Thermo Scientific Chemicals
$70.67 - $206.36
Chemical Identifiers
CAS | 10421-85-9 |
---|---|
Molecular Formula | C8H7ClO3 |
Molecular Weight (g/mol) | 186.59 |
MDL Number | MFCD00084962 |
InChI Key | RWOLDZZTBNYTMS-UHFFFAOYNA-N |
Synonym | 2-chloromandelic acid, 2-2-chlorophenyl-2-hydroxyacetic acid, dl-2-chloromandelic acid, 2-chlorophenyl hydroxy acetic acid, dl-2-chloro mandelic acid, 2-2-chlorophenyl-2-hydroxy-acetic acid, 2-chlorophenyl glycolic acid, 2-chloromandelicacid, 2-chloro-mandelic acid, pubchem15895 |
PubChem CID | 97720 |
IUPAC Name | 2-(2-chlorophenyl)-2-hydroxyacetic acid |
SMILES | OC(C(O)=O)C1=CC=CC=C1Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAB2152506
|
Thermo Scientific Chemicals
B2152506 |
5 g |
Each for $70.67
|
|
||||||
AAB2152514
|
Thermo Scientific Chemicals
B2152514 |
25 g |
Each for $206.36
|
|
||||||
Description
2-Chloromandelic acid, is used as a pharmaceutical intermediate as well as in the medicine industry. It is used as a intermediate of clopidrogrel.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Chloromandelic acid, is used as a pharmaceutical intermediate as well as in the medicine industry. It is used as a intermediate of clopidrogrel.
Solubility
Very soluble in water.
Notes
Light sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from acids, acid chlorides, acid anhydrides, strong oxidizing agents.
Chemical Identifiers
10421-85-9 | |
186.59 | |
RWOLDZZTBNYTMS-UHFFFAOYNA-N | |
97720 | |
OC(C(O)=O)C1=CC=CC=C1Cl |
C8H7ClO3 | |
MFCD00084962 | |
2-chloromandelic acid, 2-2-chlorophenyl-2-hydroxyacetic acid, dl-2-chloromandelic acid, 2-chlorophenyl hydroxy acetic acid, dl-2-chloro mandelic acid, 2-2-chlorophenyl-2-hydroxy-acetic acid, 2-chlorophenyl glycolic acid, 2-chloromandelicacid, 2-chloro-mandelic acid, pubchem15895 | |
2-(2-chlorophenyl)-2-hydroxyacetic acid |
Specifications
80°C to 85°C | |
5 g | |
MFCD00084962 | |
2-chloromandelic acid, 2-2-chlorophenyl-2-hydroxyacetic acid, dl-2-chloromandelic acid, 2-chlorophenyl hydroxy acetic acid, dl-2-chloro mandelic acid, 2-2-chlorophenyl-2-hydroxy-acetic acid, 2-chlorophenyl glycolic acid, 2-chloromandelicacid, 2-chloro-mandelic acid, pubchem15895 | |
RWOLDZZTBNYTMS-UHFFFAOYNA-N | |
2-(2-chlorophenyl)-2-hydroxyacetic acid | |
97720 | |
98% |
10421-85-9 | |
C8H7ClO3 | |
2367070 | |
Very soluble in water. | |
OC(C(O)=O)C1=CC=CC=C1Cl | |
186.59 | |
186.6 | |
2-Chloromandelic acid |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 233-900-9
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only