Learn More
2-Chlorophenol, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 180990050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Chlorophenol | |
95-57-8 | |
100.0 | |
1.2620g/mL | |
63°C | |
Glass bottle | |
1.5580 to 1.5600 | |
MFCD00002159 | |
06,183 | |
15,2157 | |
Solubility in water: 28.5g/l (20°C). Other solubilities: freely soluble in solutions of sodium hydroxide,, alcohol and ether, soluble in aromatic solvents | |
C1=CC=C(C(=C1)O)Cl | |
128.56 | |
4.11 mPa.s (25°C) | |
128.56 |
98+% | |
98.0 | |
8.0°C | |
173.0°C to 175.0°C | |
98% min. (GC) | |
C6H5ClO | |
ClC6H4OH | |
5 g | |
1.262 | |
o-chlorophenol, phenol, 2-chloro, 2-hydroxychlorobenzene, o-chlorphenol, o-chlorophenic acid, phenol, o-chloro, 2-chloro-1-hydroxybenzene, chlorophenol, septi-kleen, 2-chloro-phenol | |
ISPYQTSUDJAMAB-UHFFFAOYSA-N | |
2-chlorophenol | |
7245 | |
CHEBI:47083 | |
98+% |
Chemical Identifiers
95-57-8 | |
128.56 | |
ISPYQTSUDJAMAB-UHFFFAOYSA-N | |
7245 | |
2-chlorophenol |
C6H5ClO | |
MFCD00002159 | |
o-chlorophenol, phenol, 2-chloro, 2-hydroxychlorobenzene, o-chlorphenol, o-chlorophenic acid, phenol, o-chloro, 2-chloro-1-hydroxybenzene, chlorophenol, septi-kleen, 2-chloro-phenol | |
CHEBI:47083 | |
C1=CC=C(C(=C1)O)Cl |
Safety and Handling
GHS H Statement
Toxic to aquatic life with long lasting effects.
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for severa
GHS Signal Word: Warning
EINECSNumber : 202-433-2
RTECSNumber : SK2625000
TSCA : TSCA
RUO – Research Use Only