Learn More
2-Chlorophenyl cyclopentyl ketone, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0644818
Description
2-Chlorophenyl cyclopentyl ketone is used as a reagent in the preparation of bicyclic and polycyclic aromatic hydrocarbons. It acts as a pharmaceutical intermediate and also as an intermediate of ketamine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Chlorophenyl cyclopentyl ketone is used as a reagent in the preparation of bicyclic and polycyclic aromatic hydrocarbons. It acts as a pharmaceutical intermediate and also as an intermediate of ketamine.
Solubility
Slightly miscible with chloroform and ethyl acetate.
Notes
Incompatible with oxidizing agents.
Specifications
2-Chlorophenyl cyclopentyl ketone | |
1.16 | |
>100°C (212°F) | |
C12H13ClO | |
MFCD00038367 | |
1873022 | |
Slightly miscible with chloroform and ethyl acetate. | |
C1CCC(C1)C(=O)C2=CC=CC=C2Cl | |
208.685 | |
208.69 |
6740-85-8 | |
130°C (5 mmHg) | |
Aromatic | |
1.547 | |
50 g | |
2-chlorophenyl cyclopentyl ketone, 2-chlorophenyl cyclopentyl methanone, o-chlorophenyl cyclopentyl ketone, o-chlorobenzoylcyclopentane, 2-chlorophenyl-cyclopentylketone, methanone, 2-chlorophenyl cyclopentyl, 2-chlorobenzoylcyclopentane, acmc-1b5a4, 0-chlorphenylcyclopentylketone, 1-2-chlorobenzoyl cyclopentane | |
QIJMMRNZBJHXRI-UHFFFAOYSA-N | |
(2-chlorophenyl)-cyclopentylmethanone | |
81223 | |
97% |
Chemical Identifiers
6740-85-8 | |
208.685 | |
QIJMMRNZBJHXRI-UHFFFAOYSA-N | |
81223 | |
C1CCC(C1)C(=O)C2=CC=CC=C2Cl |
C12H13ClO | |
MFCD00038367 | |
2-chlorophenyl cyclopentyl ketone, 2-chlorophenyl cyclopentyl methanone, o-chlorophenyl cyclopentyl ketone, o-chlorobenzoylcyclopentane, 2-chlorophenyl-cyclopentylketone, methanone, 2-chlorophenyl cyclopentyl, 2-chlorobenzoylcyclopentane, acmc-1b5a4, 0-chlorphenylcyclopentylketone, 1-2-chlorobenzoyl cyclopentane | |
(2-chlorophenyl)-cyclopentylmethanone |
Safety and Handling
EINECSNumber : 229-802-0
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only