missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloropyridine-3-boronic acid, 96%, Thermo Scientific Chemicals

$71.77 - $597.16

Chemical Identifiers

CAS 381248-04-0
Molecular Formula C5H5BClNO2
Molecular Weight (g/mol) 157.36
MDL Number MFCD03094997
InChI Key VRDAOVQZVXYRNH-UHFFFAOYSA-N
Synonym 2-chloropyridine-3-boronic acid, 2-chloropyridin-3-yl boronic acid, 2-chloro-3-pyridylboronic acid, 2-chloro-3-pyridineboronic acid, 2-chloropyridine-3-boronicacid, 2-chloropyridin-3-yl-3-boronic acid, 2-chloro-3-pyridyl boronic acid, boronic acid, 2-chloro-3-pyridinyl, pubchem5078
PubChem CID 2762704
IUPAC Name (2-chloropyridin-3-yl)boronic acid
SMILES OB(O)C1=CC=CN=C1Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL2030303
View Documents
Thermo Scientific Chemicals
L2030303
1 g
Each for $71.77
 
AAL2030306
View Documents
Thermo Scientific Chemicals
L2030306
5 g
Each for $597.16
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

381248-04-0
157.36
VRDAOVQZVXYRNH-UHFFFAOYSA-N
2762704
OB(O)C1=CC=CN=C1Cl
C5H5BClNO2
MFCD03094997
2-chloropyridine-3-boronic acid, 2-chloropyridin-3-yl boronic acid, 2-chloro-3-pyridylboronic acid, 2-chloro-3-pyridineboronic acid, 2-chloropyridine-3-boronicacid, 2-chloropyridin-3-yl-3-boronic acid, 2-chloro-3-pyridyl boronic acid, boronic acid, 2-chloro-3-pyridinyl, pubchem5078
(2-chloropyridin-3-yl)boronic acid

Specifications

381248-04-0
C5H5BClNO2
1 g
2-chloropyridine-3-boronic acid, 2-chloropyridin-3-yl boronic acid, 2-chloro-3-pyridylboronic acid, 2-chloro-3-pyridineboronic acid, 2-chloropyridine-3-boronicacid, 2-chloropyridin-3-yl-3-boronic acid, 2-chloro-3-pyridyl boronic acid, boronic acid, 2-chloro-3-pyridinyl, pubchem5078
OB(O)C1=CC=CN=C1Cl
157.36
157.37
2-Chloropyridine-3-boronic acid
160°C
MFCD03094997
9119913
VRDAOVQZVXYRNH-UHFFFAOYSA-N
(2-chloropyridin-3-yl)boronic acid
2762704
96%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only