missing translation for 'onlineSavingsMsg'
Learn More

2-chloropyrimidine-5-carboxaldehyde, 97%

CAS: 933702-55-7 | C5H3ClN2O | 142.54 g/mol

$487.27 - $487.27

Chemical Identifiers

CAS 933702-55-7
Molecular Formula C5H3ClN2O
Molecular Weight (g/mol) 142.54
MDL Number MFCD10696891
InChI Key LJYQVOPFBNMTKJ-UHFFFAOYSA-N
Synonym 2-chloropyrimidine-5-carboxyaldehyde, 2-chloropyrimidine-5-carboxaldehyde, 2-chloro-5-pyrimidinecarboxaldehyde, pubchem23896, 2-chloro-5-formyl pyrimidine, chloropyrimidine-5-carbaldehyde, 2-chloro-pyrimidine-5-carbaldehyde, 2-chloranylpyrimidine-5-carbaldehyde, 5-pyrimidinecarboxaldehyde, 2-chloro
PubChem CID 21698350
IUPAC Name 2-chloropyrimidine-5-carbaldehyde
SMILES ClC1=NC=C(C=O)C=N1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC438450010
View Documents
Thermo Scientific Chemicals
438450010
1 g Glass bottle
Each for $487.27
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

933702-55-7
142.54
LJYQVOPFBNMTKJ-UHFFFAOYSA-N
21698350
ClC1=NC=C(C=O)C=N1
C5H3ClN2O
MFCD10696891
2-chloropyrimidine-5-carboxyaldehyde, 2-chloropyrimidine-5-carboxaldehyde, 2-chloro-5-pyrimidinecarboxaldehyde, pubchem23896, 2-chloro-5-formyl pyrimidine, chloropyrimidine-5-carbaldehyde, 2-chloro-pyrimidine-5-carbaldehyde, 2-chloranylpyrimidine-5-carbaldehyde, 5-pyrimidinecarboxaldehyde, 2-chloro
2-chloropyrimidine-5-carbaldehyde

Specifications

933702-55-7
100.0
White to Yellow
96% min. (GC)
C5H3ClN2O
1 g
LJYQVOPFBNMTKJ-UHFFFAOYSA-N
2-chloropyrimidine-5-carbaldehyde
21698350
97%
2-Chloropyrimidine-5-carboxaldehyde
96.0
113°C to 118°C
Authentic
Glass bottle
MFCD10696891
2-chloropyrimidine-5-carboxyaldehyde, 2-chloropyrimidine-5-carboxaldehyde, 2-chloro-5-pyrimidinecarboxaldehyde, pubchem23896, 2-chloro-5-formyl pyrimidine, chloropyrimidine-5-carbaldehyde, 2-chloro-pyrimidine-5-carbaldehyde, 2-chloranylpyrimidine-5-carbaldehyde, 5-pyrimidinecarboxaldehyde, 2-chloro
ClC1=NC=C(C=O)C=N1
142.54
142.55
Crystalline Powder or Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unw

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only