Learn More
2-chloropyrimidine-5-sulfonyl chloride, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 429630010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Chloropyrimidine-5-sulfonyl chloride | |
96.0 | |
64.0°C to 67.0°C | |
Authentic | |
Glass bottle | |
MFCD06657208 | |
2-chloro-pyrimidine-5-sulfonyl chloride, 2-chloropyrimidine-5-sulfonylchloride, 2-chloro-5-pyrimidinesulfonyl chloride, 5-pyrimidinesulfonyl chloride, 2-chloro, 2-chloro-pyrimidine-5-sulfonylchloride | |
ClC1=NC=C(C=N1)S(Cl)(=O)=O | |
213.03 | |
213.04 | |
Crystalline Powder or Powder |
98026-88-1 | |
100.0 | |
White to Cream | |
96% min. (GC) | |
C4H2Cl2N2O2S | |
1 g | |
DMAOAJYJIATWJS-UHFFFAOYSA-N | |
2-chloropyrimidine-5-sulfonyl chloride | |
43244366 | |
97% |
Chemical Identifiers
98026-88-1 | |
213.03 | |
DMAOAJYJIATWJS-UHFFFAOYSA-N | |
43244366 | |
ClC1=NC=C(C=N1)S(Cl)(=O)=O |
C4H2Cl2N2O2S | |
MFCD06657208 | |
2-chloro-pyrimidine-5-sulfonyl chloride, 2-chloropyrimidine-5-sulfonylchloride, 2-chloro-5-pyrimidinesulfonyl chloride, 5-pyrimidinesulfonyl chloride, 2-chloro, 2-chloro-pyrimidine-5-sulfonylchloride | |
2-chloropyrimidine-5-sulfonyl chloride |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
RUO – Research Use Only