missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Cyanobenzoic acid, 94%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 398140010

 View more versions of this product

Catalog No. AC398140010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2-Cyanobenzoic acid
92.0
Authentic
Glass bottle
NCC6H4CO2H
1g
DTNSDCJFTHMDAK-UHFFFAOYSA-N
2-cyanobenzoic acid
138061
94%
3839-22-3
100.0
92% min. (HPLC)
C8H5NO2
MFCD00045796
o-cyanobenzoic acid, cyanobenzoic acid, benzoic acid, 2-cyano, 2-carboxybenzonitrile, 2-carboxy-benzonitrile, 2-cyanobenzoicacid, 2-cyano-benzoic acid, pubchem12513, acmc-1bn2v
C1=CC=C(C(=C1)C#N)C(=O)O
147.13
147.13

Chemical Identifiers

3839-22-3
147.13
DTNSDCJFTHMDAK-UHFFFAOYSA-N
138061
C1=CC=C(C(=C1)C#N)C(=O)O
C8H5NO2
MFCD00045796
o-cyanobenzoic acid, cyanobenzoic acid, benzoic acid, 2-cyano, 2-carboxybenzonitrile, 2-carboxy-benzonitrile, 2-cyanobenzoicacid, 2-cyano-benzoic acid, pubchem12513, acmc-1bn2v
2-cyanobenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only