Learn More
2-cyanoethyl N,n-diisopropylchlorophosphoramidite, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 374830050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Cyanoethyl N,N-diisopropylchlorophosphoramidite | |
96.0 | |
White to Yellow | |
110°C | |
97% | |
C9H18ClN2OP | |
[(CH3)2CH]2NP(Cl)O(CH2)2CN | |
5 g | |
2-cyanoethyl n,n-diisopropylchlorophosphoramidite, 2-cyanoethyl-n,n-diisopropylchlorophosphoramidite, phosphoramidochloridous acid, bis 1-methylethyl-, 2-cyanoethyl ester, 2-cyanoethoxy diisopropylamino chlorophosphine, 3-chloro diisopropylamino phosphino oxy propanenitrile, 3-chloro-diisopropylamino phosphanyl oxypropanenitrile, 3-chloro diisopropylamino phosphanyl oxy propanenitrile, acmc-20anxd, 2-cyanoethyl diisopropylchloro-phosphoramidite, 2-cyanoethyl diisopropylphosphoramidochloridite | |
QWTBDIBOOIAZEF-UHFFFAOYSA-N | |
3-[chloro-[di(propan-2-yl)amino]phosphanyl]oxypropanenitrile | |
2734844 | |
97% |
89992-70-1 | |
100.0 | |
1.0610g/mL | |
Authentic | |
Glass bottle | |
1.4760 to 1.4800 | |
MFCD00011544 | |
1.061 | |
Solubility in water: reacts. | |
CC(C)N(C(C)C)P(OCCC#N)Cl | |
236.68 | |
236.68 | |
Crystalline Powder |
Chemical Identifiers
89992-70-1 | |
236.68 | |
QWTBDIBOOIAZEF-UHFFFAOYSA-N | |
2734844 | |
CC(C)N(C(C)C)P(OCCC#N)Cl |
C9H18ClN2OP | |
MFCD00011544 | |
2-cyanoethyl n,n-diisopropylchlorophosphoramidite, 2-cyanoethyl-n,n-diisopropylchlorophosphoramidite, phosphoramidochloridous acid, bis 1-methylethyl-, 2-cyanoethyl ester, 2-cyanoethoxy diisopropylamino chlorophosphine, 3-chloro diisopropylamino phosphino oxy propanenitrile, 3-chloro-diisopropylamino phosphanyl oxypropanenitrile, 3-chloro diisopropylamino phosphanyl oxy propanenitrile, acmc-20anxd, 2-cyanoethyl diisopropylchloro-phosphoramidite, 2-cyanoethyl diisopropylphosphoramidochloridite | |
3-[chloro-[di(propan-2-yl)amino]phosphanyl]oxypropanenitrile |
Safety and Handling
GHS H Statement
Catches fire spontaneously if exposed to air.
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Handle under inert gas.
Protect from moisture.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Immediately call a PO
GHS Signal Word: Danger
Recommended Storage : May develop turbidity on storage
RUO – Research Use Only