missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Cyclohexylcyclohexanone 97.0+%, TCI America™
$152.86 - $452.97
Chemical Identifiers
| CAS | 90-42-6 |
|---|---|
| Molecular Formula | C12H20O |
| Molecular Weight (g/mol) | 180.29 |
| MDL Number | MFCD00013795 |
| InChI Key | UOBQDYFTAJKQAL-UHFFFAOYNA-N |
| PubChem CID | 7016 |
| IUPAC Name | [1,1'-bi(cyclohexane)]-2-one |
| SMILES | O=C1CCCCC1C1CCCCC1 |
Chemical Identifiers
| 90-42-6 | |
| 180.29 | |
| UOBQDYFTAJKQAL-UHFFFAOYNA-N | |
| [1,1'-bi(cyclohexane)]-2-one |
| C12H20O | |
| MFCD00013795 | |
| 7016 | |
| O=C1CCCCC1C1CCCCC1 |
Specifications
| 90-42-6 | |
| 269°C | |
| MFCD00013795 | |
| UOBQDYFTAJKQAL-UHFFFAOYNA-N | |
| [1,1'-bi(cyclohexane)]-2-one | |
| 7016 | |
| ≥97.0% (GC) | |
| 2-Cyclohexylcyclohexanone |
| Yellow | |
| C12H20O | |
| 5 g | |
| O=C1CCCCC1C1CCCCC1 | |
| 180.29 | |
| 180.29 | |
| Liquid |
Safety and Handling
RTECSNumber : DT7400000
TSCA : Yes