missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Decyn-1-ol, 97%, Thermo Scientific Chemicals

$111.88 - $361.89

Chemical Identifiers

CAS 4117-14-0
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.253
MDL Number MFCD00041567
InChI Key GFLHGTKUCYPXHB-UHFFFAOYSA-N
Synonym 2-decyn-1-ol, 2-decyne-1-ol, n-heptylhydroxymethylacetylene, unii-l4949b9xh9, decynol, pubchem11035, acmc-1arrx, heptyl hydroxymethyl acetylene, 2-decyn-1-ol,
PubChem CID 77763
IUPAC Name dec-2-yn-1-ol
SMILES CCCCCCCC#CCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0552306
View Documents
Thermo Scientific Chemicals
L0552306
5 g
Each for $111.88
 
AAL0552314
View Documents
Thermo Scientific Chemicals
L0552314
25 g
Each for $361.89
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4117-14-0
154.253
GFLHGTKUCYPXHB-UHFFFAOYSA-N
77763
CCCCCCCC#CCO
C10H18O
MFCD00041567
2-decyn-1-ol, 2-decyne-1-ol, n-heptylhydroxymethylacetylene, unii-l4949b9xh9, decynol, pubchem11035, acmc-1arrx, heptyl hydroxymethyl acetylene, 2-decyn-1-ol,
dec-2-yn-1-ol

Specifications

4117-14-0
114°C to 116°C (6 mmHg)
C10H18O
MFCD00041567
1852842
GFLHGTKUCYPXHB-UHFFFAOYSA-N
dec-2-yn-1-ol
77763
97%
0.855
>110°C (230°F)
1.4585
5 g
2-decyn-1-ol, 2-decyne-1-ol, n-heptylhydroxymethylacetylene, unii-l4949b9xh9, decynol, pubchem11035, acmc-1arrx, heptyl hydroxymethyl acetylene, 2-decyn-1-ol,
CCCCCCCC#CCO
154.253
154.25
2-Decyn-1-ol
Safety and Handling

Safety and Handling

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only