missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2'-Deoxy-5-fluorouridine 98.0+%, TCI America™
$154.05 - $801.86
Chemical Identifiers
CAS | 50-91-9 |
---|---|
Molecular Formula | C9H11FN2O5 |
Molecular Weight (g/mol) | 246.19 |
MDL Number | MFCD00006530 |
InChI Key | ODKNJVUHOIMIIZ-RRKCRQDMSA-N |
Synonym | floxuridine, 2'-deoxy-5-fluorouridine, 5-fluoro-2'-deoxyuridine, 5-fluorodeoxyuridine, fluorodeoxyuridine, floxuridin, fudr, deoxyfluorouridine, fluoruridine deoxyribose, fdurd |
PubChem CID | 5790 |
ChEBI | CHEBI:60761 |
IUPAC Name | 5-fluoro-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione |
SMILES | OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(F)C(=O)NC1=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
D2235100MG
|
TCI America
D2235100MG |
100 mg |
N/A
|
|
||||||
D2235500MG
|
TCI America
D2235500MG |
500 mg |
N/A
|
|
||||||
D22351G
|
TCI America
D22351G |
1 g |
N/A
|
|
||||||
Chemical Identifiers
50-91-9 | |
246.19 | |
ODKNJVUHOIMIIZ-RRKCRQDMSA-N | |
5790 | |
5-fluoro-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione |
C9H11FN2O5 | |
MFCD00006530 | |
floxuridine, 2'-deoxy-5-fluorouridine, 5-fluoro-2'-deoxyuridine, 5-fluorodeoxyuridine, fluorodeoxyuridine, floxuridin, fudr, deoxyfluorouridine, fluoruridine deoxyribose, fdurd | |
CHEBI:60761 | |
OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(F)C(=O)NC1=O |
Specifications
50-91-9 | |
White | |
MFCD00006530 | |
2811 | |
ODKNJVUHOIMIIZ-RRKCRQDMSA-N | |
5-fluoro-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione | |
5790 | |
246.19 | |
Crystalline Powder |
150°C | |
C9H11FN2O5 | |
100 mg | |
floxuridine, 2'-deoxy-5-fluorouridine, 5-fluoro-2'-deoxyuridine, 5-fluorodeoxyuridine, fluorodeoxyuridine, floxuridin, fudr, deoxyfluorouridine, fluoruridine deoxyribose, fdurd | |
OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(F)C(=O)NC1=O | |
246.19 | |
CHEBI:60761 | |
≥98.0% (HPLC,T) | |
2′-Deoxy-5-fluorouridine |
Safety and Handling
RTECSNumber : YU7525000
TSCA : No