missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Deoxy-D-glucose 97.0+%, TCI America™
$81.32 - $695.51
Chemical Identifiers
CAS | 154-17-6 |
---|---|
Molecular Formula | C6H12O5 |
Molecular Weight (g/mol) | 164.16 |
MDL Number | MFCD00151328 |
InChI Key | PMMURAAUARKVCB-PHUJZJCSNA-N |
Synonym | deoxyglucose, 2-deoxy-d-mannose, 2-deoxy-d-arabinohexose, unii-9g2mp84a8w, d-arabino-hexose, 2-deoxy, arabino-hexose, 2-deoxy, d-glucose, 2-deoxy, 3r,4s,5r-3,4,5,6-tetrahydroxyhexanal, 2 deoxyglucose, 2 deoxy d glucose |
PubChem CID | 17751002 |
IUPAC Name | (4R,5S,6R)-6-(hydroxymethyl)oxane-2,4,5-triol |
SMILES | OC[C@H]1OC(O)C[C@@H](O)[C@@H]1O |
Chemical Identifiers
154-17-6 | |
164.16 | |
PMMURAAUARKVCB-PHUJZJCSNA-N | |
17751002 | |
OC[C@H]1OC(O)C[C@@H](O)[C@@H]1O |
C6H12O5 | |
MFCD00151328 | |
deoxyglucose, 2-deoxy-d-mannose, 2-deoxy-d-arabinohexose, unii-9g2mp84a8w, d-arabino-hexose, 2-deoxy, arabino-hexose, 2-deoxy, d-glucose, 2-deoxy, 3r,4s,5r-3,4,5,6-tetrahydroxyhexanal, 2 deoxyglucose, 2 deoxy d glucose | |
(4R,5S,6R)-6-(hydroxymethyl)oxane-2,4,5-triol |
Specifications
154-17-6 | |
C6H12O5 | |
1 g | |
PMMURAAUARKVCB-PHUJZJCSNA-N | |
(4R,5S,6R)-6-(hydroxymethyl)oxane-2,4,5-triol | |
17751002 | |
≥97.0% (GC) | |
2-Deoxy-D-glucose |
150°C | |
MFCD00151328 | |
deoxyglucose, 2-deoxy-d-mannose, 2-deoxy-d-arabinohexose, unii-9g2mp84a8w, d-arabino-hexose, 2-deoxy, arabino-hexose, 2-deoxy, d-glucose, 2-deoxy, 3r,4s,5r-3,4,5,6-tetrahydroxyhexanal, 2 deoxyglucose, 2 deoxy d glucose | |
OC[C@H]1OC(O)C[C@@H](O)[C@@H]1O | |
164.16 | |
164.16 | |
Crystalline Powder |
Safety and Handling
RTECSNumber : MQ3325000
TSCA : Yes