missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific Chemicals 2'-Deoxyadenosine hydrate, 99%
$995.34 - $995.34
Chemical Identifiers
CAS | 16373-93-6 |
---|---|
Molecular Formula | C10H13N5O3 |
Molecular Weight (g/mol) | 251.25 |
MDL Number | MFCD00149364 |
InChI Key | OLXZPDWKRNYJJZ-YUZWJPFSNA-N |
Synonym | 2'-deoxyadenosine monohydrate, 2r,3s,5r-5-6-amino-9h-purin-9-yl-2-hydroxymethyl tetrahydrofuran-3-ol hydrate, adenosine, 2'-deoxy-, monohydrate, adenosine, 2'-deoxy-, monohydrate, 2-deoxyadenosine hydrate, deoxyadenosine hydrate, deoxyadenosine hydrate, a-d-ribofuranosyl adenine, deoxyadenosine monohydrate, deoxyadenosine monohydrate, 2-deoxyadenosine monohydrate |
PubChem CID | 9549172 |
SMILES | NC1=C2N=CN([C@H]3C[C@H](O)[C@@H](CO)O3)C2=NC=N1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC154990250
|
Thermo Scientific Chemicals
154990250 |
25 g | Glass bottle |
Each for $995.34
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
16373-93-6 | |
251.25 | |
OLXZPDWKRNYJJZ-YUZWJPFSNA-N | |
9549172 |
C10H13N5O3 | |
MFCD00149364 | |
2'-deoxyadenosine monohydrate, 2r,3s,5r-5-6-amino-9h-purin-9-yl-2-hydroxymethyl tetrahydrofuran-3-ol hydrate, adenosine, 2'-deoxy-, monohydrate, adenosine, 2'-deoxy-, monohydrate, 2-deoxyadenosine hydrate, deoxyadenosine hydrate, deoxyadenosine hydrate, a-d-ribofuranosyl adenine, deoxyadenosine monohydrate, deoxyadenosine monohydrate, 2-deoxyadenosine monohydrate | |
NC1=C2N=CN([C@H]3C[C@H](O)[C@@H](CO)O3)C2=NC=N1 |
Specifications
16373-93-6 | |
100.0 | |
White to Beige | |
98.5% min. (HPLC) | |
C10H13N5O3 | |
25 g | |
Solubility in water: 0.3% in water (20°C). Other solubilities: soluble in dmf,formamide,hot dioxane,methanol,ethanol,insoluble in hydrocarbons | |
NC1=C2N=CN([C@H]3C[C@H](O)[C@@H](CO)O3)C2=NC=N1 | |
251.25 | |
251.24 | |
Crystalline Powder |
98.5 | |
185.0°C to 187.0°C | |
Authentic | |
Glass bottle | |
MFCD00149364 | |
2'-deoxyadenosine monohydrate, 2r,3s,5r-5-6-amino-9h-purin-9-yl-2-hydroxymethyl tetrahydrofuran-3-ol hydrate, adenosine, 2'-deoxy-, monohydrate, adenosine, 2'-deoxy-, monohydrate, 2-deoxyadenosine hydrate, deoxyadenosine hydrate, deoxyadenosine hydrate, a-d-ribofuranosyl adenine, deoxyadenosine monohydrate, deoxyadenosine monohydrate, 2-deoxyadenosine monohydrate | |
OLXZPDWKRNYJJZ-YUZWJPFSNA-N | |
5-(6-amino-9H-purin-9-yl)-2-(hydroxymethyl)oxolan-3-ol | |
9549172 | |
99% | |
2′-Deoxyadenosine hydrate |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
RUO – Research Use Only