missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals 2'-Deoxyadenosine monohydrate, 99%

Supplier:  Thermo Scientific Chemicals A1116614

 View more versions of this product

Catalog No. AAA1116614


Explore available promotions

Description

Description

2?-Deoxyadenosineise is used by some cells as an energy source under energy stress conditions and to affect cAMP levels. 2?-dAdo is used in comparison studies of the functions of adenosine analogues on various biological processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Deoxyadenosineise is used by some cells as an energy source under energy stress conditions and to affect cAMP levels. 2′-dAdo is used in comparison studies of the functions of adenosine analogues on various biological processes.

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications

Specifications

2'-Deoxyadenosine monohydrate
186°C to 189°C
MFCD00149364
5191174
Soluble in water.
−25° (c=0.5 water)
(2R,3S,5R)-5-(6-aminopurin-9-yl)-2-(hydroxymethyl)oxolan-3-ol;hydrate
9549172
99%
16373-93-6
C10H13N5O3
25 g
2'-deoxyadenosine monohydrate, 2?-deoxyadenosine monohydrate, adenosine, 2'-deoxy-, monohydrate, adenosine, 2'-deoxy-, monohydrate, 2'-deoxyadenosine hydrate, 2-deoxyadenosine hydrate, 9-2-deoxy-, a-d-ribofuranosyl adenine, a-d-ribofuranosyl adenine, deoxyadenosine monohydrate, 2-deoxyadenosine monohydrate
OLXZPDWKRNYJJZ-YUZWJPFSNA-N
NC1=C2N=CN([C@H]3C[C@H](O)[C@@H](CO)O3)C2=NC=N1
251.25
269.26 (251.24 Anhydrous)

Chemical Identifiers

16373-93-6
251.25
OLXZPDWKRNYJJZ-YUZWJPFSNA-N
9549172
NC1=C2N=CN([C@H]3C[C@H](O)[C@@H](CO)O3)C2=NC=N1
C10H13N5O3
MFCD00149364
2'-deoxyadenosine monohydrate, 2?-deoxyadenosine monohydrate, adenosine, 2'-deoxy-, monohydrate, adenosine, 2'-deoxy-, monohydrate, 2'-deoxyadenosine hydrate, 2-deoxyadenosine hydrate, 9-2-deoxy-, a-d-ribofuranosyl adenine, a-d-ribofuranosyl adenine, deoxyadenosine monohydrate, 2-deoxyadenosine monohydrate
(2R,3S,5R)-5-(6-aminopurin-9-yl)-2-(hydroxymethyl)oxolan-3-ol;hydrate
Videos
Safety and Handling

Safety and Handling

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 213-488-7

RTECSNumber : AU7358600

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only