Learn More
Thermo Scientific Chemicals 2'-Deoxyguanosine hydrate, 99%
$52.60 - $52.60
Chemical Identifiers
CAS | 961-07-9 |
---|---|
Molecular Formula | C10H13N5O4 |
Molecular Weight (g/mol) | 267.25 |
MDL Number | MFCD00150760 |
InChI Key | YKBGVTZYEHREMT-DWFKHXNSNA-N |
Synonym | 2'-deoxyguanosine, deoxyguanosine, guanine deoxyriboside, 2-deoxyguanosine, guanosine, 2'-deoxy, guanine deoxy nucleoside, 2'-deoxy-guanosine, 2;-deoxyguanosine, unii-g9481n71ro, guanine, 9-2-deoxy-beta-d-erythro-pentofuranosyl |
PubChem CID | 187790 |
ChEBI | CHEBI:17172 |
IUPAC Name | 2-amino-9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one |
SMILES | NC1=NC2=C(N=CN2[C@H]2C[C@H](O)[C@@H](CO)O2)C(=O)N1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL14519MC
|
Thermo Scientific Chemicals
L14519MC |
100 mg |
Each for $52.60
|
|
|||||
Description
Deoxyguanosine (dG) is a purine nucleoside that upon sequential phosphoylation (kinases) forms dGTP which is used by DNA polymerases and reverse transcriptases to synthesis DNA(s). Deoxyguanosine is the most electron rich of the four canonical bases and includes many nucleophilic sites which are susceptible to oxidative damage. This makes deoxyguanosine and its oxidized derivatives useful reagents to study mechanisms of oxidative damage to nucleosides and nucleotides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDeoxyguanosine (dG) is a purine nucleoside that upon sequential phosphoylation (kinases) forms dGTP which is used by DNA polymerases and reverse transcriptases to synthesis DNA(s). Deoxyguanosine is the most electron rich of the four canonical bases and includes many nucleophilic sites which are susceptible to oxidative damage. This makes deoxyguanosine and its oxidized derivatives useful reagents to study mechanisms of oxidative damage to nucleosides and nucleotides.
Solubility
Slightly soluble in water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
961-07-9 | |
267.25 | |
YKBGVTZYEHREMT-DWFKHXNSNA-N | |
187790 | |
2-amino-9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one |
C10H13N5O4 | |
MFCD00150760 | |
2'-deoxyguanosine, deoxyguanosine, guanine deoxyriboside, 2-deoxyguanosine, guanosine, 2'-deoxy, guanine deoxy nucleoside, 2'-deoxy-guanosine, 2;-deoxyguanosine, unii-g9481n71ro, guanine, 9-2-deoxy-beta-d-erythro-pentofuranosyl | |
CHEBI:17172 | |
NC1=NC2=C(N=CN2[C@H]2C[C@H](O)[C@@H](CO)O2)C(=O)N1 |
Specifications
961-07-9 | |
MFCD00150760 | |
39814 | |
Slightly soluble in water. | |
−32° (c=0.5 in Water) | |
2-amino-9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one | |
187790 | |
267.25 (Anhydrous) | |
2'-Deoxyguanosine hydrate |
C10H13N5O4 | |
100 mg | |
2'-deoxyguanosine, deoxyguanosine, guanine deoxyriboside, 2-deoxyguanosine, guanosine, 2'-deoxy, guanine deoxy nucleoside, 2'-deoxy-guanosine, 2;-deoxyguanosine, unii-g9481n71ro, guanine, 9-2-deoxy-beta-d-erythro-pentofuranosyl | |
YKBGVTZYEHREMT-DWFKHXNSNA-N | |
NC1=NC2=C(N=CN2[C@H]2C[C@H](O)[C@@H](CO)O2)C(=O)N1 | |
267.25 | |
CHEBI:17172 | |
99% |
Safety and Handling
EINECSNumber : 213-505-8
RTECSNumber : MF8760000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only