missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2'-Deoxyinosine, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals J6141114

 View more versions of this product

Catalog No. AAJ6141114


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2'-Deoxyinosine
>250°C
C10H12N4O4
25 g
2'-deoxyinosine, deoxyinosine, inosine, 2'-deoxy, 2-deoxyinosine, 9-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-9h-purin-6-ol, unii-hn0rq6sbwq, delta-ino, d-ino, hn0rq6sbwq, 9-2-deoxy-beta-d-ribofuranosyl hypoxanthine
OC[C@H]1O[C@H](C[C@@H]1O)N1C=NC2=C1N=CNC2=O
252.23
CHEBI:28997
Crystalline Powder
890-38-0
White
MFCD00005762
33517
VGONTNSXDCQUGY-YUZWJPFSNA-N
9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
65058
252.23

Chemical Identifiers

890-38-0
252.23
VGONTNSXDCQUGY-YUZWJPFSNA-N
65058
9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
C10H12N4O4
MFCD00005762
2'-deoxyinosine, deoxyinosine, inosine, 2'-deoxy, 2-deoxyinosine, 9-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-9h-purin-6-ol, unii-hn0rq6sbwq, delta-ino, d-ino, hn0rq6sbwq, 9-2-deoxy-beta-d-ribofuranosyl hypoxanthine
CHEBI:28997
OC[C@H]1O[C@H](C[C@@H]1O)N1C=NC2=C1N=CNC2=O
Safety and Handling

Safety and Handling

EINECSNumber : 212-964-1

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only