Learn More
2-(Diethylamino)ethyl chloride hydrochloride, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A151070E
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-(Diethylamino)ethyl chloride hydrochloride | |
206°C to 210°C | |
C6H15Cl2N | |
MFCD00012519 | |
UN2928 | |
Moisture sensitive | |
RAGSWDIQBBZLLL-UHFFFAOYSA-N | |
2-chloro-N,N-diethylethanamine;hydrochloride | |
13363 | |
≥98% |
869-24-9 | |
5 | |
(CH3CH2)2NCH2CH2Cl·HCl | |
2500 g | |
3591991 | |
2-chloro-n,n-diethylethanamine hydrochloride, 2-diethylaminoethylchloride hydrochloride, 2-chlorotriethylamine hydrochloride, 2-chloro-n,n-diethylethylamine hydrochloride, 2-diethylamino ethyl chloride hydrochloride, diethylaminoethyl chloride hydrochloride, n,n-diethyl-2-chloroethylamine hydrochloride, 2-chloroethyl diethylamine hydrochloride, unii-ycy6eqt3wa, ethanamine, 2-chloro-n,n-diethyl-, hydrochloride | |
[H+].[Cl-].CCN(CC)CCCl | |
172.09 | |
172.1 |
Chemical Identifiers
869-24-9 | |
172.09 | |
RAGSWDIQBBZLLL-UHFFFAOYSA-N | |
13363 |
C6H15Cl2N | |
MFCD00012519 | |
2-chloro-n,n-diethylethanamine hydrochloride, 2-diethylaminoethylchloride hydrochloride, 2-chlorotriethylamine hydrochloride, 2-chloro-n,n-diethylethylamine hydrochloride, 2-diethylamino ethyl chloride hydrochloride, diethylaminoethyl chloride hydrochloride, n,n-diethyl-2-chloroethylamine hydrochloride, 2-chloroethyl diethylamine hydrochloride, unii-ycy6eqt3wa, ethanamine, 2-chloro-n,n-diethyl-, hydrochloride | |
[H+].[Cl-].CCN(CC)CCCl |
Safety and Handling
GHS H Statement
H300-H310-H330-H314-H318-H341
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of causing genetic defects.
P260-P262-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H300+H310+H330-H314-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, CORROSIVE, ORGANIC, N.O.S.
EINECSNumber : 212-786-4
RTECSNumber : YE1050000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only